Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ACR5000GS

Millipore

GST Reagent Kit

suitable for protein purification

Synonym(s):

Protein purification kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NA.25

product line

Amicon®

technique(s)

protein purification: suitable (GSH-tagged proteins)

shipped in

wet ice

General description

For use with the Amicon Pro Affinity Conc: 3 ml GST Bind resin, 125 mg Glutathione (reduced, free acid), 25ml GST Bind/Wash Buffer (10X), 5 ml GST Reconstitution buffer (10X)
GST Reagent Kit for GSH-tagged protein purification.

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

Flash Point(F)

95.9 °F

Flash Point(C)

35.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Protein purification is simpler with Amicon Pro. The device delivers consistent sample preparation for more reliable recovery, uncompromised purity and easier data generation.

Protocols

If an antibody must be purified from a small sample volume (0.5-10 mL), then the Amicon® Pro Purification System is recommended.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service