Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ACK5030GS

Millipore

Amicon® Pro Affinity Concentration Kit GST with 30kDa Amicon® Ultra-0.5 Device

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101

material

Amicon Ultra styrene-butadiene device
Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding)
polyethylene cap
polyethylene frit
polypropylene device (for exchange)
polypropylene frit
polypropylene tube (AU0.5; for collection)
polypropylene tube (for collection)
polypropylene tube (holder)
regenerated cellulose membrane

sterility

non-sterile

manufacturer/tradename

Amicon® Pro

parameter

10 mL sample volume

technique(s)

buffer exchange: suitable
centrifugation: suitable
protein purification: suitable (antibody labeling, concentration, enrichment)

L × diam.

12 cm (4.7 in.) × 2.97 cm

filtration area

1 cm2

working volume

10 mL

pore size

30 kDa NMWCO

Related Categories

General description

PLCGC

Application

Amicon Pro Affinity Concentration Kit GST - A Centrifugal Tool for purifying & concentrating GSH-tagged proteins.
Ultrafiltration

Features and Benefits

Minimum Final Concentrate Volume: 15 μL

Components

This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO 30 kDa, 3 ml GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/ml agarose), 125 mg Glutathione (reduced, free acid), 25ml GST Bind/Wash Buffer (10X), 5 ml GST Reconstitution buffer (10X)

Physical form

Color Code:Clear with pink cap

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoon Sun Chun et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-08)
Accumulation of β-amyloid (Aβ) in the brain has been implicated in the pathology of Alzheimer's disease (AD). Aβ is produced from the Aβ precursor protein (APP) through the amyloidogenic pathway by β-, and γ-secretase. Alternatively, APP can be cleaved by
Jocelyne Hanquier et al.
Experimental parasitology, 211, 107868-107868 (2020-03-03)
Toxoplasma gondii is a protozoan parasite of great importance in human and veterinary health. The frontline treatment of antifolates suffers a variety of drawbacks, including toxicity and allergic reactions, underscoring the need to identify novel drug targets for new therapeutics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service