Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABT49

Sigma-Aldrich

Anti-Dynamin-2 Antibody

serum, from rabbit

Synonym(s):

dynamin 2, dynamin II, dynamin-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$467.00

$467.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$467.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$467.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DNM2(1785)

General description

Dynamin-2 is a ubiquitously expressed, membrane associated protein belonging to the Dynamin protein family of GTPases, a group consisting of three isoforms with distinct tissue specificity and function. Dynamin localizes to plasma membrane-coated pits by its interactions with amphiphysin through the dynamin PRD domain. Dynamin-2 is a component in the manufacture of microtubule bundles, and is capable of both binding and hydrolyzing GTP. It is also likely to be involved in the trafficking mechanisms for vesicles, especially those pertaining to endocytosis. Defects in the expression of Dynamin-2 have been shown to cause Charcot-Marie-Tooth disease dominant intermediate type B (CMTDIB), and centronuclear myopathy autosomal dominant (ADCNM). There is also some interest in Dynamin-2 as a potential regulator for adenovirus targeting cancer therapies.

Immunogen

KLH-conjugated linear peptide corresponding to human Dynamin-2.

Application

Detect Dynamin-2 using this Anti-Dynamin-2 Antibody validated for use in WB.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1:1,000 dilution of the antibody detected Dynamin-2 in 10 µg of HeLa cell lysate.

Target description

~ 98 kDa observed

Physical form

Rabbit polyclonal serum containing 0.5% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service