Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABT47

Sigma-Aldrich

Anti-Claudin-7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Claudin-7, CLDN-7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CLDN7(1366)

General description

Claudin-7 is a member of a family of claudins that are tight junction integral membrane proteins with four transmembrane domains and two extracellular loops. Claudin-7 has been shown to be under-expressed in invasive ductal carcinomas and colorectal carcinomas as compared to normal epithelial cells. Claudin-7 differs from other claudins because it has both structural and regulatory functions, regulating the expression of prostate specific antigen.

Immunogen

KLH-conjugated linear peptide corresponding to human Claudin-7.

Application

Anti-Claudin-7 Antibody is an antibody against Claudin-7 for use in WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Western Blot in BALB3T3/A31 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Claudin-7 in 10 µg of BALB3T3/A31 cell lysate.

Target description

~22 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
BALB3T3/A31 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service