Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABT342

Sigma-Aldrich

Anti-ADAP Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

ADAP Antibody, Adhesion and degranulation adapter protein, FYN binding protein, FYB-120, 130, SLAP-130, FYN-T-binding protein, SLP-76-associated phosphoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FYB(2533)

General description

The adhesion and degranulation adaptor protein (ADAP) acts as an adapter protein of the FYN and LCP2 signaling cascades in T-cells. ADAP modulates the expression of interleukin-2 (IL-2) and it is suggested to be involved in platelet activation. Additionally ADAP is shown to prevent the degradation of SKAP1 and SKAP2 and may play a role in linking T-cell signaling to the remodeling of the actin cytoskeleton. ADAP is expressed in hematopoietic tissues such as myeloid and T-cells, spleen and thymus. At least two isoforms of ADAP are known to exist.

Specificity

This antibody recognizes the C-terminus of ADAP.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human ADAP.

Application

Detect ADAP using this rabbit polyclonal antibody, Anti-ADAP Antibody validated for use in western blotting & ICC.
Immunocytochemistry Analysis: 10 µg/mL from a representative lot detected ADAP in K562 cells.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected ADAP in 15 µg of K562 cell lysate.

Target description

~130 kDa observed. Uncharacterized bands may appear in some lysates. Uniprot describes 2 isoforms at ~85 kDa and ~91 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service