Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABT272

Sigma-Aldrich

Anti-SUN2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

SUN domain-containing protein 2, Protein unc-84 homolog B, Rab5-interacting protein, Rab5IP, Sad1/unc-84 protein-like 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$532.00

$532.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$532.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$532.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SUN2(25777)

General description

SUN domain-containing protein 2 (SUN2), or Protein UNC-84 homolog B (UNC84B), is a single-pass nuclear envelope transmembrane protein responsible for nuclear migrations that are essential for development. SUN2 has a predicted transmembrane domain and a C-terminal region with similarity to the S. pombe spindle pole body protein Sad1. SUN2 interacts with RAB5A and is widely expressed, and highly expressed in the heart, lungs and muscles. UNC-84 may function to facilitate a nuclear-centrosomal interaction required for nuclear migration and anchorage.

Immunogen

Epitope: N-terminus
Linear peptide corresponding to the N-terminus of human Sun2.

Application

Detect SUN2 using this Anti-SUN2 antibody validated for use in western blotting & ICC.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
Western Blotting Analysis: A 1:500 dilution from a representative lot detected SUN2 in 10 µg of A431, Jurkat, L6, and NIH/3T3 cell lysate.
Western Blotting Analysis: A representative lot detected SUN2 recombinant protein (Haque, F., et al. (2010). Journal of Biol. Chem. 285(5): 3487-3498).
Immunocytochemistry Analysis: A representative lot detected SUN2 in human foreskin fibroblasts and NIH/3T3 cells (Haque, F., et al. (2010). Journal of Biol. Chem. 285(5): 3487-3498).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected SUN2 in 10 µg of HEK293 cell lysate.

Target description

~ 78 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service