Skip to Content
MilliporeSigma
All Photos(2)

Documents

ABT21

Sigma-Aldrich

Anti-Amphiphysin II Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

bridging integrator 1, amphiphysin-like, box dependant MYC interacting protein 1, Amphiphysin II, Box-dependent myc-interacting protein 1, Amphiphysin-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AMPH(273)

General description

Amphiphysin is a nerve terminal-enriched protein thought to be involved in synaptic vesicle endocytosis. One of three amphiphysin homologs, amphiphysin II may interact with amphiphysin I in order to regulate synaptic vesicle endocytosis. During clathrin-mediated endocytosis, amphiphysin has been reported to bind to a component of the clathrin coat, clathrin assembly protein 2. Mutations in the expression of amphiphysin II may be causal to defects in the endocytosis pathway. Expression of amphiphysin II has been observed in neurons along with some endocrine cell types and spermatocytes.

Specificity

The antibody is raised against human amphiphysin II, but recognizes both amphiphysin I and Amphiphysin II (see Molecular Weight Notes).

Immunogen

Epitope: Unknown
KLH-conjugated recombinant protein corresponding to human Amphiphysin II.

Application

Detect Amphiphysin II using this Anti-Amphiphysin II Antibody validated for use in WB.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling
Western Blot Analysis: 1:1,000 dilution from a previus lot detected Amphiphysin II in 10 µg of rat brain and mouse brain membrane tissue lysates.

Quality

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected Amphiphysin II in 10 µg of mouse brain tissue lysate.

Target description

It is well established (Ramjaun, 1997) that both Amphiphysin I and II run substantially slower on SDS-PAGE with a larger apparent molecular weight than the predicted molecular weights.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service