Skip to Content
MilliporeSigma
All Photos(2)

Documents

ABT189

Sigma-Aldrich

Anti-Tropomyosin 4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Tropomyosin alpha-4 chain, TM30p1, Tropomyosin-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TPM4(7171)

General description

Tropomyosin-4 is also called Tropomyosin alpha-4 chain and TM30p1. Tropomyosin-4 binds calcium and to actin filaments in muscle and non-muscle cells. Tropomyosin-4 associates with the troponin complex, and plays a role in the regulation of smooth and striated muscle contraction, as well as the stabilization of cytoskeleton actin filaments in non-muscle cells. Tropomyosin-4 is found in cardiac tissue and platelets.

Specificity

This antibody recognizes the C-terminus of Tropomyosin 4.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human Tropomyosin 4.

Application

Anti-Tropomyosin 4 Antibody is an antibody against Tropomyosin 4 for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected Tropomyosin 4 in human prostate cancer sections and human heart sections tissue.

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: A 1:2,500 dilution of this antibody detected Tropomyosin 4 in 10 µg of NIH/3T3 cell lysate.

Target description

~33 kDa observed. The calculated molecular weight is 29 kDa, however Tropomyosin 4 has been shown as a ~33 kDa band in western blots (Brooke, K., et al. (2006). Bone. 39:697-705).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service