Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS94

Sigma-Aldrich

Anti-MLK1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Mitogen-activated protein kinase kinase kinase 9, Mixed lineage kinase 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$357.00

$357.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$357.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$357.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAP3K9(4293)

General description

MLK1 (MAP3K9) is a serine–threonine kinase of the Mixed-Lineage Kinase (MLK) family capable of activating the c-Jun N-terminal Kinase (JNK)/Mitogen-Activated Protein Kinase (MAPK) pathway and regulating the other two prinicpal MAPK cascades, p38 and extracellular signal regulated kinase (ERK). Immunoblots and mass spectrometry show MLK is threonine phosphorylated in or near the activation loop. Studies show that MLKs play a role in the TGF-beta1-induced cell death mechanism.

Immunogen

KLH-conjugated linear peptide corresponding to human MLK1.

Application

Anti-MLK1 Antibody detects level of MLK1 & has been published & validated for use in IH(P).
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

Quality

Evaluated by Immunohistochemistry in colorectal adenocarcinoma tissue.

Immunohistochemistry Analysis: A 1:300 dilution of this antibody detected MLK1 in colorectal adenocarcinoma tissue.

Target description

121 kDa (isoform 1), 123 kDa (isoform 2) calculated

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Colorectal adenocarcinoma tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service