Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ABS787

Sigma-Aldrich

Anti-PGK1 Antibody (Center)

from rabbit

Synonym(s):

PGK1, PGKA, Phosphoglycerate kinase 1, Cell migration-inducing gene 10 protein, Primer recognition protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

Gene Information

human ... PGK1(5230)

General description

Phosphoglycerate kinase 1 (PGK1) and encoded by the gene names Pgk1 or Pgk-1, is important for phosphoprotein glycolysis. PGK1 is an ATP-generating glycolytic enzyme that forms part of the glycolytic pathway that is often highly expressed in prostate cancer and is regulated by hypoxia-inducible factor-1α. Extracellular PGK1 facilitates the cleavage of plasminogen generating the vascular inhibitor angiostatin.

Immunogen

Epitope: Central region
KLH-conjugated synthetic peptide corresponding to the central region of human PGK1.

Application

Detect PGK1 using this rabbit polyclonal Anti-PGK1 Antibody, Cat. No. ABS787, validated for use in Western Blotting, Immunohistochemistry, Immunocytochemistry, and Flow Cytometry.
Research Category
Metabolism
Research Sub Category
Glucose/Glycogen Metabolism
Western Blotting Analysis (WB): A 1:1000 dilution from a representative lot of this antibody detected PGK1 in multiple lysates. See chart for reactivity detail.

Immunohistochemistry Analysis (IHC): A 1:10-50 dilution from a representative lot detected PGK1 in human cancer tissue.

Immunocytochemistry Analysis (ICC): A 1:10-50 dilution from a representative lot detected PGK1 in HeLa cells.

Flow Cytometry (FC): A 1:10-50 dilution of this antibody detected PGK1 in HeLa cells.

Quality

Evaluated by Western Blotting in HT1376 cell lysate.

Western Blotting Analysis (WB): A 1:1000 dilution of this antibody detected PGK1 in HT1376 cell lysate.

Target description

~48 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Ammonium Sulfate
Format: Purified
Purified rabbit polyclonal in buffer containing PBS with 0.09% (W/V) sodium azide. This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Penelope J Boyd et al.
PLoS genetics, 13(4), e1006744-e1006744 (2017-04-21)
Degeneration and loss of lower motor neurons is the major pathological hallmark of spinal muscular atrophy (SMA), resulting from low levels of ubiquitously-expressed survival motor neuron (SMN) protein. One remarkable, yet unresolved, feature of SMA is that not all motor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service