Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS564

Sigma-Aldrich

Anti-Serpinf2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Alpha-2-antiplasmin, Alpha-2-AP, Alpha-2-plasmin inhibitor, Alpha-2-PI, Serpin F2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SERPINF2(5345)

General description

Serpin F2, also known as Alpha-2-antiplasmin Alpha-2-AP, Alpha-2-plasmin inhibitor, Alpha-2-PI, and encoded by the gene name SERPINF2, AAP, or PLI, is a member of the Serpin family. Serpin F2 is a serine protease inhibitor. As an inhibitor, the major targets of Serpin F2 are plasmin and trypsin, but it also inactivates matriptase-3/TMPRSS7 and chymotrypsin. Serpin F2 forms protease inhibiting heterodimer with TMPRSS7. Serpin F2 is expressed by the liver and secreted in plasma. Mutations in Serpin F2 have been associated with Alpha-2-plasmin inhibitor deficiency (APLID), an autosomal recessive disorder resulting in severe hemorrhagic diathesis.

Immunogen

Epitope: Alpha-2-antiplasmin mature chain
KLH-conjugated linear peptide corresponding to the Alpha-2-antiplasmin mature chain of human Serpinf2.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Serpinf2 in human liver and human liver cancer tissue.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-Serpinf2 antibody is validated for use in WB, IH for the detection of Serpinf2.

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Serpinf2 in 10 µg of human liver tissue lysate.

Target description

~73 kDa observed. Predicated to work with both glycos and non-glycos isoforms.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service