Skip to Content
MilliporeSigma
All Photos(1)

Documents

ABS547

Sigma-Aldrich

Anti-Pancreatic Lipase/PNLIP Antibody

serum, from rabbit

Synonym(s):

Pancreatic triacylglycerol lipase, PL, PTL, Pancreatic lipa, Pancreatic Lipase/PNLIP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

neutralization: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PNLIP(5406)

General description

Pancreatic triacylglycerol lipase (EC 3.1.1.3; UniProt P35638; also known as Pancreatic lipase, PL, PTL, Triacylglycerol acylhydrolase) is encoded by the PNLIP (Also known as PNLIPD) gene in human (Gene ID 5406). PTL is a pancreas synthesized carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides (TAGs). The dietary TAGs are partially digested by the gastric lipase in the stomach. The emulsion particles then mix with digestive enzymes from the pancreas in the duodenum where digestion proceeds. The pancreas secretes multiple lipases, including pancreatic triglyceride lipase (PTL), carboxyl ester lipase (CEL), and pancreatic lipase-related protein-1 and -2 (PLRP1 & PLRP2). Unlike PLRP2, PLRP1 exhibits little activity against triglycerides and no measurable activity against phospholipids, galactolipids or cholesterol esters. Experimental evidences suggest that PLRP1 plays a negative regulatory role against PLT–colipase-mediated dietary triglyceride digestion. Pancreas-derived lipases are essential for the efficient digestion of dietary TAG. Pancreatic insufficiency or exocrine pancreas failure results in steatorrhea or steatorrhoea, where increased fat excretion is seen in fecal samples.

Specificity

Reacts with pancreatic triglyceride lipase (PTL), pancreatic lipase-related protein-1 & -2 (PLRP1 & PLRP2), and the truncated PLRP2 W340X mutant.

Immunogen

Recombinant protein corresponding to human Pancreatic Lipase/PNLIP.

Application

Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-Pancreatic Lipase/PNLIP Antibody is validated for use in Western Blotting, Neutralizing for the detection of Pancreatic Lipase/PNLIP .
Western Blotting Analysis: A representative lot detected recombinant mouse pancreatic triglyceride lipase (PTL/PNLIP), as well as endogenous pancreatic lipase-related protein-2 (PLRP2) in mouse pancreas (Xiao, X., et al. (2011) J. of Lipid Res. 52(5):982-990).
Western Blotting Analysis: A representative lot detected both wild-type and W340X mutated recombinant human pancreatic lipase-related protein-2 (PLRP2) in culture media and lysates from transfected 293T and COS-7 cells (Xiao, X., et al. (2011) J. Biol. Chem. 286(30):26353-26363).
Neutralizing Analysis: A representative lot inhibited colipase-dependent pancreatic lipase-related protein-2 (PLRP2) enzymatic activity in pancreas extracts from 4-day old mice (D′Agostino D, and Lowe ME. (2004) J. Nutr. 134(1):132-134).

Quality

Evaluated by Western Blotting in human pancreas tissue lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected Pancreatic Lipase/PNLIP in 10 µg of human pancreas tissue lysate.

Target description

~53 kDa observed. 51.1 kDa (Human PTL, calculated), 51.4 kDa (Mouse & Rat PTL, calculated), 51.8/51.9 kDa (Human PLRP1/2, calculated), 52.7/52.6 kDa (Mouse PLRP1/2, calculated), 52.4/52.5 kDa (Rat PLRP1/2, calculated). Target band may migrate as a doublet on gel due to glycosylation.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service