Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABS521

Sigma-Aldrich

Anti-RSK4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Q9UK32, KS6A6_HUMAN, RPS6KA6, RSK4, Q9UK32, B2R854, Q6FHX2, Q8WX28, Q9H4S6, Ribosomal protein S6 kinase alpha-6, p90RSK6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$468.00

$468.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$468.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$468.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

human (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RPS6KA6(27330)

General description

RSK1, RSK2, RSK3, and RSK4 (ribosomal protein S6 kinase, 90 kDa, polypeptide 1,2,3, and 4; S6K-alpha 1,2,3, and 4) are Ser/Thr kinases which are activated by MAP Kinases. Ribosomal S6-kinase 4 (RSK4) is thought to be mapped in the critical region that is commonly deleted in X-linked deafness type 3, mental retardation, and choroideremia. It may be involved in normal neuronal development. RSK4 is also thought to play a role in non-growth factor signaling and may also be important in tumor suppression.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of Human RSK4.

Application

Immunoprecipitation analysis: 10 ug of Cat. No. ABS521, Anti-RSK4 was used to immunoprecipitate RSK4 in 0.5 mg PC12 or EGF treated PC12 cell lysate
Immunoprecipitation analysis: 5 ug of Cat. No. ABS521, Anti-RSK4 was used to immunoprecipitate RSK4 in 0.665mg of PC12 cell lysate. Uncharacterized bands may be detected at high antibody concentration or longer exposures
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

General Post-translation Modification
This Anti-RSK4 Antibody is validated for use in Western Blotting and Immunoprecipitation for the detection of RSK4.

Quality

Evaluated by Western Blotting in rat PC12 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected RSK4 in 10 µg of rat PC12 cell lysate.

Target description

~83 kDa observed. Uncharacterized band(s) may appear in some lysates.

Linkage

Replaces: 04-1132

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service