Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS500

Sigma-Aldrich

Anti-CDT2 Antibody

serum, from rabbit

Synonym(s):

Denticleless protein homolog, B1- and CUL4-associated factor 2, Lethal(2) denticleless protein homolog, Retinoic acid-regulated nuclear matrix-associated protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$415.00

$415.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$415.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$415.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DTL(51514)

General description

CDT2, also known as DTL, Denticleless protein homolog, DDB1- and CUL4-associated factor 2, Lethal(2) denticleless protein homolog, or Retinoic acid-regulated nuclear matrix-associated protein, and encoded by the gene DTL/CDT2/CDW1/DCAF2/L2DTL/RAMP, is a component of the DCX(DTL) E3 ubiquitin ligase complex and is the substrate-specific adapter for the DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control, DNA damage response and translesion DNA synthesis. The DCX(DTL) complex, also named CRL4(CDT2) complex, mediates the polyubiquitination and subsequent degradation of CDT1 and CDKN1A/p21(CIP1). CDT2 is important because degradation of CDT1 in response to DNA damage is necessary to ensure proper cell cycle regulation of DNA replication. CDT2 is localized to the nucleus primarily but it translocates to the cytoplasm during mitosis. CDT2 is expressed in multiple tissues at varying degrees with high expression in adult placenta, testis, thymus, bone marrow and fetal all fetal tissues, and low expression in adult muscle, spleen and lymph node.

Immunogen

Recombinant protein corresponding to human CDT2.

Application

Anti-CDT2 Antibody detects level of CDT2 & has been published & validated for use in CDT2.
Research Category
Signaling
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blotting Analysis: A representative lot from an independent laboratory detected CDT2 in transfected 293T cell lysates (Abbas, T., et al. (2013). Mol Cell. 49(6):1147-1158.).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CDT2 in 10 µg of A431 cell lysate.

Target description

~85 kDa observed

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neysha Tirado-Class et al.
Life science alliance, 7(1) (2023-11-09)
DDB1- and CUL4-associated factors (DCAFs) CDT2 and DCAF14 are substrate receptors for Cullin4-RING E3 ubiquitin ligase (CRL4) complexes. CDT2 is responsible for PCNA-coupled proteolysis of substrates CDT1, p21, and SET8 during S-phase of cell cycle. DCAF14 functions at stalled replication
Debra L Evans et al.
Cell cycle (Georgetown, Tex.), 15(1), 95-105 (2016-01-16)
The timely and precise duplication of cellular DNA is essential for maintaining genome integrity and is thus tightly-regulated. During mitosis and G1, the Origin Recognition Complex (ORC) binds to future replication origins, coordinating with multiple factors to load the minichromosome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service