Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS459

Sigma-Aldrich

Anti-USP10 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ubiquitin carboxyl-terminal hydrolase 10, Deubiquitinating enzyme 10, Ubiquitin thioesterase 10, Ubiquitin-specific-processing protease 10, UPS10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... USP10(9100)

General description

Ubiquitin carboxyl-terminal hydrolase 10 (USP10), also known as Ubiquitin-specific-processing protease 10, belongs to the peptidase C19 family and functions to catalyze the cleavage of ubiquitin from ubiquitin-conjugated protein substrates such as p53/TP53, SNX3 and CFTR. USP10 is widely expressed and is down-regulated in renal cell carcinomas.

Immunogen

KLH-conjugated linear peptide corresponding to human USP10.

Application

Anti-USP10 Antibody is a highly specific rabbit polyclonal antibody, that targets Ubiquitin carboxyl-terminal hydrolase 10 & has been tested in western blotting & ICC.
Immunocytochemistry Analysis: 20 µg/mL from a representative lot detected USP10 in Jurkat cells.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected USP10 in 15 µg of Jurkat cell lysate.

Target description

~122 kDa observed. Uniprot describes 3 isoforms at ~~87 kDa, ~88 kDa, and ~93 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service