Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS456

Sigma-Aldrich

Anti-WNT10A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein Wnt-10a, Wingless-type MMTV integration site family 10a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

shipped in

wet ice

Gene Information

human ... WNT10A(80326)

General description

Wingless-type MMTV integration site family 10a (WNT10A) plays crucial role in normal development such as regulation of cell fate, patterning during embryogenesis and neoplastic transformation. Elevated levels of WNT10A have been detected in several human tumors and cancer cell lines including colorectal cancer and chronic lymphocytic leukemia. This evidence suggests that uncontrolled Wnt signaling may contribute to the abnormal apoptosis that characterizes these malignancies. Defects in WNT10A have been shown to be related and possibly are the cause of ectodermal dysplasia anhidrotic (EDA), also known ectodermal dysplasia hypohidrotic autosomal recessive (HED). Other recent evidence had suggested that mutation in WNT10A is associated with odonto-onycho-dermal dysplasia (OODD), an autosomal recessive ectodermal dysplasia and Schopf-Schulz-Passarge syndrome (SSPS).

Immunogen

KLH-conjugated linear peptide corresponding to human WNT10A.

Application

Anti-WNT10A Antibody is a highly specific rabbit polyclonal antibody, that targets Wnt & has been tested in western blotting, IHC (Paraffin) & Immunofluorescence.
Immunohistochemistry Analysis: 10 µg/mL from a representative lot detected WNT10A in human skeletal muscle tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected WNT10A in human skeletal muscle cells.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in RAW264.7 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected WNT10A in 15 µg of RAW264.7 cell lysate.

Target description

~46 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
RAW264.7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service