Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS448

Sigma-Aldrich

Anti-CSN8 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

CSN8 Antibody, COP9 signalosome complex subunit 8, SGN8, JAB1-containing signalosome subunit 8, CSN8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$350.00

$350.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$350.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$350.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... COPS8(10920)

General description

COP9 signalosome complex subunit 8 (CSN8, COPS8), also known as Signalosome subunit 8 (SGN8), is a nuclear-enriched protein complex belonging to the CSN8 family. It is comprised of eight subunits that interact with deubiquitinating enzymes and protein kinases. The CSN complex is involved in the regulation of various signaling and developmental processes, including DNA repair, cell cycle regulation, embryogenesis, axonal guidance, MAPK cascade, immune response, and hormonal signaling.

Specificity

This antibody recognizes the C-terminus of CSN8.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human CSN8.

Application

Anti-CSN8 Antibody is a highly specific rabbit polyclonal antibody, that targets COP9 signalosome complex subunit 8 & has been tested in western blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected CSN8 in 15 µg of human liver tissue lysate.

Target description

~27 and ~30 kDa observed. Uniprot describes 2 isoforms at ~18 kDa and ~23 kDa Uncharacterized bands may appear in some lysate(s).

Linkage

Replaces: MABN1028

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service