Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS446

Sigma-Aldrich

Anti-GPVI Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

GPVI Antibody, Glycoprotein VI, GPIV, Platelet glycoprotein VI precursor, GP6, Glycoprotein 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GP6(51206)

General description

Glycoprotein 6 (GP6), or glycoprotein VI (GPVI), is a member of the immunoglobulin gene superfamily, closely related to Fc receptor gamma chain (FcRgamma) and natural killer receptors. GP6 is a collagen receptor involved in collagen-induced platelet adhesion and activation, and may contribute to arterial and venous thrombus formation. GP6 is a membrane glycoprotein that plays a crucial role in platelet procoagulant activity and subsequent thrombin and fibrin formation. GP6 is expressed in megakaryocytes and platelets. The signaling pathway of GP6 involves the FcRgamma, the Src kinases (likely Fyn/Lyn), the adapter protein LAT and leads to the activation of phospholipase C gamma2. It has been shown that bleeding disorders such as Bleeding Disorder Platelet-Type 11 (BDPLT11), a mild to moderate bleeding disorder caused by defective platelet activation and aggregation in response to collagen, is caused by defects in GP6.

Specificity

This antibody recognizes the C-terminus of GPVI.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human GPVI.

Application

Anti-GPVI Antibody is a highly specific rabbit polyclonal antibody, that targets Glycoprotein 6 & has been tested in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: 10 µg/mL from a representative lot detected GPVI in human heart tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected GPVI in human heart cells.

Quality

Evaluated by Western Blotting/Peptide Inhibition Assay in A20 cell lysate.

Western Blotting/Peptide Inhibition Assay Analysis: 1 µg/mL of this antibody detected GPVI in 15 µg of A20 cell lysate.

Target description

~115 kDa observed. Uniprot describes 3 isoforms at ~35 kDa, ~37 kDa, and ~67 kDa

Linkage

Replaces: MABT473

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biswajit Saha et al.
Science advances, 7(30) (2021-07-23)
Platelets extravasate from the circulation into tumor microenvironment, enable metastasis, and confer resistance to chemotherapy in several cancers. Therefore, arresting tumor-platelet cross-talk with effective and atoxic antiplatelet agents in combination with anticancer drugs may serve as an effective cancer treatment
Benjamin E J Spurgeon et al.
Current protocols, 1(5), e112-e112 (2021-05-06)
Platelets are small blood cells that contribute to hemostasis, immunity, and inflammation. Characterization of platelet surface markers allows for differentiation of activated platelets from resting platelets, diagnosis of platelet disorders, and investigation of platelet biology and pathology. In this article

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service