Skip to Content
MilliporeSigma
All Photos(2)

Documents

ABS236

Sigma-Aldrich

Anti-Cytochrome P450 (scc) Antibody, a.a. 509-526

from rabbit, purified by affinity chromatography

Synonym(s):

Cholesterol side-chain cleavage enzyme, mitochondrial, CYPXIA1, Cholesterol desmolase, Cytochrome P450 11A1, Cytochrome P450(scc)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

As a member of the cytochrome P450 family, Cytochrome P450 Side Chain Cleavage Enzyme (scc) catalyzes the side-chain cleavage reaction of cholesterol to pregnenolone during the first step of steroid biosynthesis. It is expressed in certain organs, such as ovaries, adrenal glands, and testis. Deficiency in CYP11A1 is commonly associated with adrenal insufficiency, and with variable degrees of disorder of sex development (DSD).

Specificity

This antibody recognizes the C-terminus of Cytochrome P450 (scc), a.a. 509-526.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of rat Cytochrome P450 (scc), a.a. 509-526.

Application

Immunohistochemistry Analysis: A 1:4,000-8,000 dilution from a representative lot detected Cytochrome P450 (scc) in rat adrenal gland and rat testis tissue.
This Cytochrome P450 (scc) antibody is validated for use in WB & IHC for the detection of the Cytochrome P450 (scc) protein.

Quality

Evaluated by Western Blot in rat adrenal tissue lysate.

Western Blot Analysis: 0.02 µg/mL of this antibody detected Cytochrome P450 (scc) in 10 µg of rat adrenal tissue lysate.

Target description

~52 kDa observed. On higher percentage gels and longer exposures, 8 kDa cleavage product can be detected.

Linkage

Replaces: AB1294

Analysis Note

Control
Rat adrenal tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Guang-Yu et al.
Molecular reproduction and development, 83(3), 226-235 (2016-03-21)
Myeloid cell leukemia-1 (MCL1), an anti-apoptotic member of the BCL2 family, is expressed abundantly in the testis. Previous characterization revealed that MCL1 is expressed exclusively in the Leydig cells in the mouse testis, yet what it does in these cells
A Berthon et al.
Human molecular genetics, 26(18), 3495-3507 (2017-09-16)
Inactivating mutations in the Armadillo repeat-containing 5 (ARMC5) gene have recently been discovered in primary macronodular adrenal hyperplasia (PMAH), a cause of Cushing syndrome. Biallelic ARMC5 inactivation in PMAH suggested that ARMC5 may have tumor suppressor functions in the adrenal
Sheu-Ran Choi et al.
Frontiers in pharmacology, 10, 1439-1439 (2019-12-24)
Research indicates that neurosteroids are locally synthesized in the central nervous system and play an important modulatory role in nociception. While the neurosteroidogenic enzyme, cytochrome P450 side-chain cleavage enzyme (P450scc), is the initiating enzyme of steroidogenesis, P450scc has not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service