Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS2270

Sigma-Aldrich

Anti-CDK11

Synonym(s):

Cell division cycle 2-like protein kinase 2, Cell division protein kinase 11A, Cyclin-dependent kinase 11A, EC:2.7.11.22, Galactosyltransferase-associated protein kinase p58/GTA, PITSLRE serine/threonine-protein kinase CDC2L

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$232.00

$232.00


Estimated to ship onApril 01, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
$232.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$232.00


Estimated to ship onApril 01, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

rabbit #5, polyclonal

mol wt

calculated mol wt 91.36 kDa
observed mol wt ~91 kDa

species reactivity

human

species reactivity (predicted by homology)

mouse

packaging

antibody small pack of 100

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

human ... CDK11A(728642)

Specificity

This rabbit polyclonal antibody detects Cyclin-dependent kinase 11A (CDK11) and targets an epitope within the N-terminal region.

Immunogen

His-tagged recombinant fragment corresponding to the first 226 amino acids from the N-terminal region of human Cyclin-dependent kinase 11A (CDK11A).

Application

Quality Control Testing

Evaluated by Western Blotting in HCT116 Cell Lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CDK11 in HCT116 cell lysate.

Tested Applications

Western Blotting Analysis: A representative lot detected Cyclin-dependent kinase 11 in Western Blotting applications (Hluchy, M., et. al. (2022). Nature. 609(7928):829-834).

ChIP-seq Anaylysis: A representative lot detected Cyclin-dependent kinase 11 in ChIP-seq applications (Hluchy, M., et. al. (2022). Nature. 609(7928):829-834).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Cyclin-dependent kinase 11A (UniProt: Q9UQ88; also known as EC:2.7.11.22, Cell division cycle 2-like protein kinase 2, Cell division protein kinase 11A, Galactosyltransferase-associated protein kinase p58/GTA, PITSLRE serine/threonine-protein kinase CDC2L2) is encoded by the CDK11A (also known as CDC2L2, CDC2L3, PITSLREB) gene (Gene ID: 728642) in human. CDK11 is a ubiquitously expressed protein that plays multiple roles in cell cycle progression, cytokinesis, and apoptosis. Eight different isoforms of CDK11 have been described that are produced by alternative splicing and alternative initiation. The p110 isoform (canonical) is suggested to be involved in pre-mRNA splicing, potentially by phosphorylating the splicing protein SFRS7. During apoptosis, it is induced by Fas or tumor necrosis factor. The p110 isoform is cleaved by caspases to produce p110C form that contains the C-terminal kinase domain and binds to the serine/threonine kinase PAK1. CDK11 is reported to associate with SF3B1 and phosphorylates threonine residues at its N terminus during spliceosome activation. This phosphorylation event is shown to be essential for the association between SF3B1 and U5 and U6 snRNAs in the activated spliceosome, termed the Bact complex. CDK11 is overexpressed in several tumor types and is a predictor of poor clinical outcome. (Ref.: Blazek, D. (2023). Clin. Transl Med. 13(3); e1201; Hluchý, M., et al. (2022). Nature. 609(7928); 829-834).

Physical form

Rabbit polyclonal antibody in serum without azide.

Reconstitution

Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service