Skip to Content
MilliporeSigma
All Photos(2)

Documents

ABS218

Sigma-Aldrich

Anti-phospho-DBC1 (Thr454) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

DBIRD complex subunit KIAA1967, Deleted in breast cancer gene 1 protein, DBC-1, DBC.1, p30 DBC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr454)

Gene Information

human ... BRINP1(1620)

General description

Deleted in breast cancer gene 1 protein (DBC1) is a large, nuclear protein with multiple functions in cell survival that binds directly to the estrogen receptor α (ERα) hormone-binding domain in a strict ligand-independent manner. DBC1 promotes p53-mediated apoptosis by binding to and inhibiting the deacetylase activity of SirT1, resulting in increased p53 acetylation levels and activity. It is ubiquitously expressed in 84 to 100% of neoplastic breast, lung, and colon tissues.

Specificity

This antibody recognizes DBC1 phosphorylated at Thr454.

Immunogen

Epitope: Phosphorylated Thr454
KLH-conjugated linear peptide corresponding to human DBC-1 phosphorylated at Thr454.

Application

Anti-phospho-DBC1 (Thr454) Antibody is an antibody against phospho-DBC1 (Thr454) for use in Western Blotting, Peptide Inhibition Assay.
Peptide Inhibition Assay: A 1:500 dilution from a representative lot blocked DBC-1 phosphorylated at Thr454 in untreated and etoposide treated Jurkat cell lysates.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blot in untreated and etoposide treated Jurkat cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected DBC-1 phosphorylated at Thr454 in 10 µg of etoposide treated Jurkat cell lysates.

Target description

~130 kDa observed. Uniprot states the calculated molecular weight is 103 kDa but has been shown at ~130kDa (Satoh, M., et al. (1995). Brain Res Dev Brain Res. 87(2):111-119).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Untreated and etoposide treated Jurkat cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service