Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS2119M

Sigma-Aldrich

Anti-TTC39B

from rabbit

Synonym(s):

Tetratricopeptide repeat protein 39B, TPR repeat protein 39B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... TTC39B(158219)

General description

Tetratricopeptide repeat protein 39B (UniProt: also known as TPR repeat protein 39B, TTC39B) is encoded by the TTC39B (also known as C9orf52) gene (Gene ID: 158219) in human. TTC39B protein contains two TPR repeats (aa 393-426 and 626-659). TCC39B protein is the product of a high density lipoprotein (HDL) gene that promotes the ubiquitination and degradation of liver X receptor (LXR). TTC39B null or deficient mice challenged with high fat/cholesterol/bile salt diet exhibit increased LXR protein and target gene expression. TTC39B deficiency is reported to stabilize LXR by reducing its polyubiquitination and proteasomal degradation. Hence, it reduces the incidence of atherosclerosis and steatohepatitis. TTC39B deficient mice display higher levels of HDL and apoplipoprotein A-1 and lower levels of hepatic triglyceride synthesis. However, it does not affect glucose tolerance or hepatic gluconeogenesis. (Ref.: Hsieh, J et al. (2016). Nature 535(7611): 303-307).

Specificity

This rabbit polyclonal antibody detects TTC39B in human and murine cells. It targets an epitope with in 20 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal half of human Tetratricopeptide repeat protein (TTC39B).

Application

Anti-TTC39B, Cat. No. ABS2119, is a rabbit polyclonal antibody that detects Tetratricopeptide repeat protein 39B and has been tested for use in Western Blotting.
Western Blotting Analysis: A representative lot detected TTC39B in cultured primary mouse hepatocytes transfected with T39 (Hsieh, J., et. al. (2016). Nature. 535(7611):303-7).

Quality

Evaluated by Western Blotting in HEK293T cells transfected with TTC39B.

Western Blotting Analysis: 1 µg/mL of this antibody detected TTC39B in 10 µg of TTC39B transfected HEK293T cell lysate.

Target description

~90 kDa observed; 76.96 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service