Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS1856

Sigma-Aldrich

Anti-PI3 Kinase Antibody, p85

from rabbit, purified by affinity chromatography

Synonym(s):

Phosphatidylinositol 3-kinase regulatory subunit alpha, PI3-kinase regulatory subunit alpha, PI3K regulatory subunit alpha, Phosphatidylinositol 3-kinase 85 kDa alpha, PtdIns-3-kinase regulatory subunit p85-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$568.00

$568.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$568.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$568.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PIK3R1(5295)

General description

Phosphatidylinositol 3-kinase regulatory subunit alpha (UniProt: Q63787; also known as PI3-kinase regulatory subunit alpha, PI3K regulatory subunit alpha, PtdIns-3-kinase regulatory subunit alpha, Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha, PI3-kinase subunit p85-alpha, PtdIns-3-kinase regulatory subunit p85-alpha) is encoded by the Pik3r1 gene (Gene ID: 25513) in rat. PI3K regulatory subunit alpha catalyzes the phosphorylation of PI(4,5)P2 inositol ring at the 3 position to generate PI(3,4,5)P3, a potent second messenger that mediates survival signaling and insulin action. PI3 Kinase is a heterodimeric complex composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. Following ligand binding to receptor and activation of receptor tyrosine kinase, the p85/p110 complex is recruited to the receptor by interaction of the SH2 domain of p85 with consensus phosphotyrosine residues on receptor tyrosine kinase. This allows the p110 catalytic subunit to come in close proximity to its lipid substrates in the cell membrane. The interaction of a receptor tyrosine kinase with the p85 subunit relieves the inhibitory effect of p85 on the p110 catalytic subunit. Activated PI 3-kinase phosphorylates phosphoinositol (PI) substrates to produce PI(3)P, PI(3,4)P2, and PI(3,4,5)P3 (PIP3). These molecules act as second messengers and recruit PI 3-kinase-dependent serine/threonine kinases (PDK1) and Akt from the cytoplasm to the plasma membrane. PI 3-kinase activity is tightly regulated in normal cells. Over-activation of this pathway can perturb the control mechanisms for cell growth and survival and contribute to metastatic competence and resistance to chemotherapy. Abnormal activation of PI 3-kinases is seen in several forms of cancer. About 30% of solid tumors contain mutations in the catalytic unit of their PI 3-kinase. PI3 kinase plays a critical role in insulin-stimulated increase in glucose uptake and glycogen synthesis in insulin-sensitive tissues. Mutations in Pik3r1 gene are known to be associated with insulin resistance.

Specificity

This polyclonal antibody detects p85 regulatory subunit of PI3 kinase in multiple species. It targets an epitope within 18 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of p85 regulatory subunit of PI-3 kinase alpha.

Application

Detect PI 3-kinase alpha, p85 subunit using this rabbit polyclonal Anti-PI3 Kinase, p85, Cat. No. ABS1856 that has been tested in Immunoprecipitation and Western Blotting.
Immunoprecipitation Analysis: 2.5 µg from a representative lot detected PI3-Kinase, p85 in jurkat cell lysate.
Research Category
Signaling

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PI3-Kinase, p85 in 10 µg of Raji cell lysate.

Target description

~80 kDa observed; 83.53 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 06-195

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinwei Wang et al.
Molecular medicine reports, 23(3) (2021-01-27)
Retinoblastoma (RB) is the most common ocular malignancy that occurs during childhood. Scavenger receptor class A member 5 (SCARA5) is considered to function as an anti‑oncogene in several types of malignant tumor. The present study investigated the functional role and underlying mechanism
Kiyomi Takase et al.
Physiological reports, 11(8), e15667-e15667 (2023-04-20)
Many kinds of fish are characterized by a limited efficiency to use carbohydrates. For this reason, raw fish and mixed feed containing a lot of fish meal have been used as feed for fish farming. However, continuing to use high-protein
Qiaoni Hu et al.
Journal of biochemical and molecular toxicology, 37(1), e23220-e23220 (2022-09-13)
We have identified that hydrogen sulfide (H2 S), a gaseous mediator, plays a crucial role in antioxidative, anti-inflammatory, and cytoprotective effects on uranium (U)-triggered rat nephrotoxicity. Pyroptosis is a special mode of inflammation and programmed cell death involved in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service