Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS1656

Sigma-Aldrich

Anti-VPS4B (UT292)

serum, from rabbit

Synonym(s):

Vacuolar protein sorting-associated protein 4B, Cell migration-inducing gene 1 protein, Protein SKD1, Suppressor of K(+) transport growth defect 1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$516.00

$516.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μL
$516.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$516.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... VPS4B(9525)

General description

Vacuolar protein sorting-associated protein 4B (EC 3.6.4.6; UniProt O75351; also known as Cell migration-inducing gene 1 protein, Protein SKD1, Suppressor of K(+) transport growth defect 1) is encoded by the VPS4B (also known as SKD1, SKD1B, VPS4-2, VPS42, MIG1) gene (Gene ID 9525) in human. SKD1 is a ubiquitously expressed protein that is involved in intracellular protein trafficking at the late endosomal protein transport step. Displays about 80% sequence homology with VPS4A. It recognizes membrane-associated ESCRT-III (endosomal sorting complex required for transport III) assemblies and catalyzes their disassembly, possibly in combination with membrane fission. Also, redistributes the ESCRT-III components to the cytoplasm for further rounds of multi-vesicular body (MVB) sorting. In conjunction with the ESCRT machinery, it also appears to function in topologically equivalent membrane fission events, such as the terminal stages of cytokinesis and enveloped virus budding (HIV-1 and other lentiviruses). VPS4A/B are required for the exosomal release of SDCBP, CD63 and syndecan. SKD1 is proposed to be monomeric or homodimeric in nucleotide-free form and to oligomerize upon binding to ATP to form two stacked hexameric or heptameric rings with a central pore through which ESCRT-III substrates are translocated in an ATP-dependent manner.

Ref.: Scheuring, S et al. (2001). J. Mol. Biol. 312, 469-480.

Specificity

VPS4B immunostaining and target band detection by UT292 was observed only in untransfected, but not VPS4B shRNA-transfected HeLa cells (Morita, E., et al. (2010). Proc. Natl. Acad. Sci. U.S.A. 107(29):12889-12894).

Immunogen

His-tagged recombinant full-length human VPS4B.

Application

Detect Vacuolar protein sorting-associated protein 4B using this rabbit polyclonal Anti-VPS4B (UT292), Cat. No. ABS1656, validated for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected VPS4B immunoreactivity concentrated at the centrosome during mitosis and at the midbody during cytokinesis in 3% paraformaldehyde-fixed HeLa cells. No staining was seen in VPS4B shRNA-transfected HeLa cells (Morita, E., et al. (2010). Proc. Natl. Acad. Sci. U.S.A. 107(29):12889-12894).

Western Blotting Analysis: A representative lot detected the VPS4B target band in lysate from untransfected, but not VPS4B shRNA-transfected HeLa cells (Morita, E., et al. (2010). Proc. Natl. Acad. Sci. U.S.A. 107(29):12889-12894).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected VPS4B in 10 µg of HEK293 cell lysate.

Target description

~52 kDa observed. 49.30 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiyu Song et al.
Cell death and differentiation, 30(9), 2151-2166 (2023-08-19)
The centrosome assembles a bipolar spindle for faithful chromosome segregation during mitosis. To prevent the inheritance of DNA damage, the DNA damage response (DDR) triggers programmed spindle multipolarity and concomitant death in mitosis through a poorly understood mechanism. We identified

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service