Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS1614

Sigma-Aldrich

Anti-Ric-8B/Synembryn-B Antibody

serum, from rabbit

Synonym(s):

Synembryn-B, Protein Ric-8B, Ric-8B, Ric-8B/Synembryn-B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$597.00

$597.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$597.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$597.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rabbit, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RIC8B(55188)

General description

Synembryn-B (UniProt Q80XE1; also known as Protein Ric-8B, Resistance to inhibitors of cholinesterase 8B) is encoded by the Ric8b (also known as BC051080) gene (Gene ID 237422) in murine species. Ric-8A and Ric-8B stimulate nucleotide exchange of all Gα subunit classes by stabilizing the Gα nucleotide-free transition state. Ric-8A acts upon Gαi/q/13, while Ric-8B targets Gαs/olf. In addition to their function as guanine nucleotide exchange factors (GEFs), Ric proteins also act as molecular chaperones that directly participate in the posttranslational folding of nascent Gα subunits to promote G-protein biosynthesis and membrane association. Ric-8 knockdown reduces Gα steady-state abundances and overexpression of Ric-8 proteins enhances cellular Gα levels. In Ric-8A– or Ric-8B–delicient ES cells, newly produced Gα subunits are defective in the initial association with an internal membrane, resulting in a ∼10-fold faster turned over rate than in WT cells.

Specificity

Expected to react with all murine and human spliced isoforms (UniProt Q80XE1 & Q9NVN3).

Immunogen

Recombinant protein corresponding to mouse Ric-8B/Synembryn-B.

Application

This Anti-Ric-8B/Synembryn-B Antibody is validated for use in Western Blotting and Immunoprecipitation for the detection of Ric-8B/Synembryn-B.
Western Blotting Analysis: A representative lot detected Ric-8B protein in murine embryonic stem (mES) cells (mESCs) derived from wild-type (WT) and Ric-8A−/− mice, but not mESCs from Ric-8B−/− mice (Gabay, M., et al. (2011). Sci Signal.4(200):ra79).
Western Blotting Analysis: A representative lot detected the endogenous Ric-8B protein in rabbit reticulocyte lysates (RRL) as well as recombinant murine Ric-8B protein (Chan, P., et al. (2013). Proc Natl Acad Sci U S A. 110(10):3794-3799).
Immunoprecipitation Analysis: A representative lot immunoprecipitated the endogenous Ric-8B protein from rabbit reticulocyte lysates/RRL (Chan, P., et al. (2013). Proc Natl Acad Sci U S A. 110(10):3794-3799).

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1:1,000 dilution of this antibody detected Ric-8B/Synembryn-B in 10 µg of Jurkat cell lysate.

Target description

~63 kDa observed. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service