Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABS1605

Sigma-Aldrich

Anti-Cytochrome p450 2J Family Antibody

serum, from rabbit

Synonym(s):

Cytochrome P450 2J, Arachidonic acid epoxygenase, CYP2J, CYPIIJ, Cytochrome P450 subfamily IIJ polypeptide, Flavoprotein-linked monooxygenase, Microsomal monooxygenase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$523.00

$523.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$523.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$523.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), sheep (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CYP2J2(1573)

General description

Cytochrome P450 2J2 (EC 1.14.14.1; UniProt P51589; also known as Arachidonic acid epoxygenase, CYPIIJ2, Cytochrome P450 subfamily IIJ polypeptide 2, Flavoprotein-linked monooxygenase, Microsomal monooxygenase) is encoded by the CYP2J2 (also known as CPJ2) gene (Gene ID 1573) in human. Cytochromes P450 (CYP) proteins are primarily membrane-associated oxidases located either in the inner membrane of mitochondria or in the endoplasmic reticulum where they function as the terminal enzymes in electron transfer chains. In addition to processing endogenous substrates, CYPs also function to metabolize exogenous drugs and potentially toxic chemicals. The human CYP superfamily consisits of 57 genes and more than 59 pseudogenes divided into 18 families and 43 subfamilies. The CYP2J subfamily of enzymes are responsible for the epoxidation of endogenous arachidonic acid (AA) into epoxyeicosatrienoic acids (EETs) and 20-hydroxyeicosatetraenoic acid (20-HETE).

Specificity

This polyclonal antiserum is also known as Anti-CYP2J2pep3. It cross-reacts with all known CYP2J subfamily P-450s, but not non-CYP2J subfamily P-450s (Yaghi, A., et al. (2003). Am. J. Physiol. Lung Cell Mol. Physiol. 285(5): L1099–L1105).

Immunogen

Epitope: Internal (C-terminal half).
Linear peptide corresponding to an internal sequence from the C-terminal half of human cytochrome p450 2J2.

Application

Anti-Cytochrome p450 2J Family Antibody is an antibody against Cytochrome p450 2J Family for use in Western Blotting
, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: An 1:8,000 dilution from a representative lot detected cardiomyocytes with overexpressed human CYP2J2 in formalin-fixed, paraffin-embedded heart tissue sections from transgenic mice carrying human CYP2J2 transgene downstream of αMHC promoter to allow cardiomyocyte-specific human CYP2J2 expression (Courtesy of Dr. Darryl C. Zeldin, NIEHS, NIH, Research Triangle Park, NC).
Western Blotting Analysis: An 1:2,000 dilution from a representative lot detected recombinant CYP2J (2J2, 2J3, 2J5, 2J8, 2J9, 2J12, 2J13), but not CYP2C (2C44, 2C50, 2C54) proteins (Courtesy of Dr. Darryl C. Zeldin, NIEHS, NIH, Research Triangle Park, NC).
Western Blotting Analysis: A representative lot detected the overexpressed human CYP2J2 in primary lung endothelial cells (ECs) from transgenic mice carrying human CYP2J2 transgene downstream of the murine Tie2 promoter to allow endothelial expression of human CYP2J2 (Deng, Y., et al. (2011). FASEB J. 25(2):703–713).
Western Blotting Analysis: A representative lot detected a 51 kDa CYP2J band in MIH-2 mouse hepatocellular carcinoma cells (HCCs), but not in C26 and MC38 mouse colon cancer cells or T3 mouse small intestine cancer cells (Homma, S., et al. (2009). Clin. Exp. Immunol. 156(2):344-352).
Western Blotting Analysis: A representative lot detected different CYP2J banding patterns between prenatal and postnatal human liver microsomal samples from human subjects ranging from 1.1 EGA (estimated gestational age) to 17-year-old of age (Gaedigk, A., et al. (2006). J. Pharmacol. Exp. Ther. 319(2): 523–532).
Western Blotting Analysis: A representative lot detected the overexpressed human CYP2J2 in heart tissues from transgenic mice carrying human CYP2J2 transgene downstream of αMHC promoter to allow cardiomyocyte-specific transgene expression (Seubert, J., et al. (2004). Circ. Res. 95(5):506-514).
Western Blotting Analysis: A representative lot detected a reduced CYP2J level in lung microsome preparations from pneumonia rats when compared with control rats (Yaghi, A., et al. (2003). Am. J. Physiol. Lung Cell Mol. Physiol. 285(5): L1099–L1105).
Western Blotting Analysis: A representative lot detected recombinant human (2J2), rat (2J3 & 2J4), and mouse (2J5, 2J6, 2J8, 2J9) CYP2J proteins (Yaghi, A., et al. (2003). Am. J. Physiol. Lung Cell Mol. Physiol. 285(5): L1099–L1105).
Immunohistochemistry Analysis: A representative lot detected the overexpressed human CYP2J2 in formalin-fixed, paraffin-embedded heart tissue sections from transgenic mice carrying human CYP2J2 transgene downstream of αMHC promoter to allow cardiomyocyte-specific transgene expression (Seubert, J., et al. (2004). Circ. Res. 95(5):506-514).
Research Category
Signaling
Research Sub Category
Enzymes & Biochemistry

Quality

Evaluated by Western Blotting in rat liver tissue lysate.

Western Blotting Analysis: An 1:5,000 dilution of this antibody detected cytochrome p450 2J protein in 10 µg of rat liver tissue lysate.

Target description

~50 kDa observed.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara A Helal et al.
Frontiers in pharmacology, 15, 1438567-1438567 (2024-08-27)
R/S enantiomers of 11-hydroxyeicosatertraenoic acid (11-HETE) are formed from arachidonic acid by enzymatic and non-enzymatic pathways. 11-HETE is predominately formed by the cytochrome P450 1B1 (CYP1B1). The role of CYP1B1 in the development of cardiovascular diseases is well established. This
K Lockhart Jamieson et al.
International journal of molecular sciences, 22(4) (2021-02-12)
Myocardial infarction (MI) accounts for a significant proportion of death and morbidity in aged individuals. The risk for MI in females increases as they enter the peri-menopausal period, generally occurring in middle-age. Cytochrome (CYP) 450 metabolizes N-3 and N-6 polyunsaturated
Yueran Li et al.
Oxidative medicine and cellular longevity, 2018, 2917981-2917981 (2018-07-27)
CYP2J proteins are present in the neural cells of human and rodent brain regions. The aim of this study was to investigate the role of brain CYP2J in Parkinson's disease. Rats received right unilateral injection with lipopolysaccharide (LPS) or 6-hydroxydopamine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service