Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS1582

Sigma-Aldrich

Anti-phospho Glucocorticoid Receptor Antibody (Ser287)

from rabbit, purified by affinity chromatography

Synonym(s):

Glucocorticoid receptor, GR, Nuclear receptor subfamily 3 group C member 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$514.00

$514.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$514.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$514.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

technique(s)

ChIP: suitable
immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer287)

Gene Information

General description

Glucocorticoid receptor (Uniprot P06536; also known as GR, Nuclear receptor subfamily 3 group C member 1) is encoded by Nr3c1 (also known as Grl, Grc) gene (Gene ID: 24413) in rat species. It has a dual mode of action whereby it acts as a transcription factor that binds to glucocorticoid response elements, both for nuclear and mitochondrial DNA, and also acts a modulator of other transcription factors. GR is composed of several conserved structural elements, including a carboxy-terminal ligand-binding domain (which contains residues critical for receptor dimerization and hormone-dependent gene transactivation), a neighboring hinge region containing nuclear localization signals, a central zinc-finger-containing DNA-binding domain, and an amino-terminal variable region that participates in ligand-independent gene transcription. In the absence of hormone, a significant population of GR is localized to the cytoplasm in an inactive form via its association with regulatory chaperone proteins, such as HSP90, HSP70, and FKBP52. Upon hormone binding, GR is released from the chaperone complex and translocates to the nucleus as a dimer to associate with glucocorticoid response elements (GREs), thereby enhancing or repressing transcription of specific target genes. GR-mediated transcriptional activation is modulated by phosphorylation and upon binding to receptor agonist it becomes hyperphosphorylated. Although basal GR phosphorylation at Ser155 and Ser287 is low, but it is highly inducible by BDNF treatment. It is been reported that BDNF-induced phosphorylation increases GR occupancy and cofactor recruitment at the promoter of a BDNF-enhanced gene.

Specificity

This polyclonal antibody detected decreased GR S287 phosphorylation in brain tissue from BDNF+/ and TrkB TrkB+/ mice, as well as GR pS287 upregulation upon inducible BDNF overexpression and Cre-dependent GR pS287 downregulation in transgenic mice with floxed GR allele (Lambert, W.M., et al. (2013). Mol. Cell. Biol. 33(18):3700-3714). The target phosphorylation site coresponds to Ser287 of rat GR spliced isoform A and Ser260 of isoform B reported by UniProt (P06536). The equivalent site is also found in human (Ser247 of isoforms Alpha-B & Beta-B, and Ser267 of all the other seven spliced isoforms except GR-P) and mouse (Ser275 of isoform 1-A & 1-B, Ser248 of isoform 2-A & 2-B) GR sequences reported by UniProt (P04150 & P06537).

Immunogen

Synthetic peptide corresponding to rat glucocorticoid receptor target site sequence containing the phosphoryated serine residue.

Application

Detect Glucocorticoid Receptor phosphorylated on Serine287 using this rabbit polyclonal Anti-GR pSer287, Cat. No. ABS1582, validated for use in Western Blotting and Immunofluorescence, Western Blotting, Western Blotting and Chromatin Immunoprecipitation (ChIP).
Western Blotting Analysis: 4.2 µg/mL from a representative lot detected 100 nM dexamethasone/100 nM of H2O2 treatment-induced glucocorticoid receptor Ser287 phosphorylation in primary ear fibroblasts from wild-type, but GR S287A-knock-in mice (Courtesy of Elina Sherestha and Michael J. Garabedian, PhD).

Chromatin Immunohistochemistry (ChIP) Analysis: A representative lot detected enhanced Ser287 phosphorylation of GR recruited to the SGK1 and GILZ regulatory regions upon dexamethasone treatment or BDNF/dexamethasone cotreatment of 293TrkB cells stably expressing wild-type rat GR (Lambert, W.M., et al. (2013). Mol. Cell. Biol. 33(18):3700-3714).

Immunofluorescence Analysis: A representative lot detection GR S287 phosphorylation in hypothalamic CRH-producing neurons by fluorescence immunohistochemistry staining of 4% paraformaldehyde-fixed free-floating mouse coronal sections. GR S287 phosphorylation decreased in brain tissue from BDNF+/ and TrkB TrkB+/ mice, increased upon inducible BDNF overexpression and decreased upon Cre-dependent excision in transgenic mice with floxed GR allele (Lambert, W.M., et al. (2013). Mol. Cell. Biol. 33(18):3700-3714).

Western Blotting Analysis: A representative lot detected BDNF-induced GR Ser287 phosphorylation in primary rat cortical neurons, PC12 cells expressing TrkB, and 293TrkB cells stably expressing wild-type rat GR, as well as upregulated GR Ser287 phosphorylation in mouse brain following dexamethasone injection or stress-induced elevation of endogenous glucocorticoids by forced swim (Lambert, W.M., et al. (2013). Mol. Cell. Biol. 33(18):3700-3714).

Quality

Evaluated by Western Blotting in mouse ear fibroblasts.

Western Blotting Analysis: 2.1 µg/mL of this antibody detected 100 nM dexamethasone/100 nM of H2O2 treatment-induced glucocorticoid receptor Ser287 phosphorylation in primary ear fibroblasts from wild-type, but GR S287A-knock-in mice.

Target description

~90 kDa observed. 85.66/81.51/85.82/81.67/64.75/60.60/82.85/78.70/82.90 kDa (Human isoform Alpha-A/Beta-A/Alpha-2; Gamma/Beta-2/GR-A alpha/GR-A beta/Alpha-B/Beta-B/hGRDelta313-338), 86.05/86.21/83.27/83.43 kDa (Mouse isoform 1-A/2-A/1-B/2-B), and 87.56/84.83 kDa (Rat isoform A/B) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified rabbit polyclonal antibody in buffer containing TBS, pH 8.0, containing no preservatives.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jordan M Buck et al.
Biochemical pharmacology, 168, 438-451 (2019-08-14)
Maternal smoking of conventional or vapor cigarettes during pregnancy, a form of developmental nicotine exposure (DNE), enhances the risk of neurodevelopmental disorders such as ADHD, autism, and schizophrenia in children. Modeling the multigenerational effects of smoking during pregnancy and nursing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service