Skip to Content
MilliporeSigma
All Photos(1)

Documents

ABS1520

Sigma-Aldrich

Anti-GRASP55 Antibody

serum, from rabbit

Synonym(s):

Golgi reassembly-stacking protein 2, GRS2, Golgi reassembly-stacking protein of 55 kDa, GRASP55, GRASP55

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

hamster, mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

GenBank accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GORASP2(26003)

General description

Golgi reassembly stacking protein of 55 kDa (also called Golgi reassembly-stacking protein of 55 kDa, GRASP55, or Grs2) is encoded by the gene designated as Gorasp2 in rat & mouse (GORASP2/GRS2/GOLPH6 in human). Golgi stack formation involves two homologous peripheral Golgi proteins, GRASP55 and GRASP65, localized to the medial-trans and cis cisternae, respectively. GRASP55 and GRASP65 stack mammalian Golgi cisternae via a common mechanism of phosphorylation-regulated oligomerization. GRASP55 stacks Golgi membranes by forming oligomers through its N-terminal GRASP domain. This process is regulated by phosphorylation within the C-terminal serine/proline-rich domain. Expression of nonphosphorylatable GRASP55 mutants enhances Golgi stacking in interphase cells and inhibits Golgi disassembly during mitosis. Knockdown of either GRASP55 or GRASP65 by siRNA reduces the number of cisternae per Golgi stack, while simultaneous knockdown of both GRASP proteins leads to disassembly of the entire stack. (PMID 20083603). Cat. No. ABS1520, Anti-GRASP55, is a rabbit polyclonal antiserum raised against a recombinant immunogen encompassing region within the C-terminal half of rat GRASP55.

Immunogen

Epitope: C-terminal
Recombinant protein corresponding to the C-terminal of rat GRASP55.

Application

Anti-GRASP55 Antibody is an antibody against GRASP55 for use in Western Blotting, Immunocytochemistry.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: A representative lot detected GRASP55 in HeLa cell lysate (Xiang, Y., and Wang, Y. (2010). J. Cell Biol. 188(2):237-251).
Western Blotting Analysis: A representative lot detected GRASP55 in CHO cells and mouse brain lysate (Joshi, G., et al (2014). PNAS. 111(13):E1230-1239).
Immunocytochemistry Analysis: A representative lot detected GRASP55 in HeLa cells (Xiang, Y., and Wang, Y. (2010). J. Cell Biol. 188(2):237-251).
Immunocytochemistry Analysis: A representative lot detected GRASP55 in CHO cells (Joshi, G., et al (2014). PNAS. 111(13):E1230-1239).

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected GRASP55 in 10 µg of mouse brain tissue lysate.

Target description

~54 kDa observed

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service