Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN898

Sigma-Aldrich

Anti-BMP2

from rabbit, purified by affinity chromatography

Synonym(s):

Bone morphogenetic protein 2, BMP-2, BMP-2A, Bone morphogenetic protein 2A

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$594.00

$594.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1489

Request a Bulk Order

Select a Size

Change View
200 μL
$594.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$594.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1489

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... BMP2(650)

General description

Bone morphogenetic protein 2 (UniProt: P12643; also known as BMP-2, BMP-2A, Bone morphogenetic protein 2A) is encoded by the BMP2 (also known as BMP2A) gene (Gene ID 650) in human. BMP2 is a member of the transforming growth factor-beta (TGF-beta) superfamily. It is synthesized as a 60-kDa precursor that is processed in the secretory pathway to a small 18-kDa monomer; 2 monomers then associate to form the active homodimer, which binds to its receptor. BMP2 can induce bone formation and regeneration during early embryonic development. It is also involved in the hedgehog pathway, TGF beta signaling pathway, and cytokine-cytokine receptor interaction. BMP2 is known to induce cartilage and bone formation and stimulates the differentiation of myoblasts into osteoblasts via the EIF2AK3-EIF2A- ATF4 pathway. BMP2 activation of EIF2AK3 stimulates phosphorylation of EIF2A, which leads to increased expression of ATF4 that plays a critical role in osteoblast differentiation.

Specificity

This rabbit polyclonal antibody targets a region present in Prepro-, pro-, and mature human BMP2.

Immunogen

GST-tagged recombinant protein composed of the propeptide C-terminal region and the mature human BMP2 N-terminal region.

Application

Anti-BMP2, Cat. No. ABN898, is a highly specific rabbit polyclonal antibody that targets bone morphogenetic protein 2 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected BMP2 immunoreactivity in human prostate cancer, cerebellum, and tonsil tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human kidney tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected BMP2 in 10 µg of human kidney tissue lysate.

Target description

~45 kDa observed. 44.70/42.48/12.90 kDa (Prepro-/pro-/mature human BMP2 monomer). Target band size appears larger due to glycosylation and dimerization. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: ABN303

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service