Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABN55

Sigma-Aldrich

Anti-Netrin-1 Antibody

serum, from rabbit

Synonym(s):

netrin 1, mouse, homolog of, Netrin 1, Ntn1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ntn1(18208)

General description

Netrin-1 is a secreted, bifunctional ligand belonging to the Netrin family; a group of proteins known to act as versatile, chemotropic axon guidance cues for CNS commissural and peripheral motor axons. Netrin-1 is necessary for spinal cord oligodendrocyte development. There are two receptors with which Netrin-1 is associated and that will cause it to be either a chemoattractant or a chemorepellant for axons. Receptor Deleted in Colorectal Cancer (DCC) regulates the attraction of axons to Netrin-1, and UNC5 mediates axon repulsion from Netrin-1. Studies have shown Netrin-1 to function as a survival factor through its relationship to its receptors, inhibiting the onset of apoptosis. Netrin-1 has also been implicated in colorectal tumorigenesis via its apoptotic regulatory role. Expression of Netrin-1 has been detected in adult axons and oligodendrocytes, and is also widely observed in developing embryonic mesodermal tissues and nervous system.

Specificity

Other homologies: Human (94% sequence homology).

Immunogen

Epitope: Unknown
Linear peptide corresponding to mouse Netrin-1.

Application

Anti-Netrin-1 Antibody detects level of Netrin-1 & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Growth Cones & Axon Guidance

Quality

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 1:10,000 dilution of this antibody detected Netrin-1 on 10 µg of mouse brain tissue lysate.

Target description

~68 kDa observed. Characteristic proteolytic fragments of Netrin-1 found in the CNS run at lower molecular weights.

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Mouse brain tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liqing Wang et al.
The Journal of comparative neurology, 525(13), 2915-2928 (2017-05-26)
Using Nogo antibodies with defined binding specificity, Nogo-B, but not Nogo-A, was localized on radial glia in the floor plate of mouse embryos. The presence of Nogo-B was confirmed in Nogo-A knockout mice. In explant cultures of embryonic day (E)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service