Skip to Content
MilliporeSigma
All Photos(1)

Documents

ABN476

Sigma-Aldrich

Anti-ACVR1C Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Activin receptor type-1C, Activin receptor type IC, ACTR-IC, Activin receptor-like kinase 7, ALK-7, ACVR1C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACVR1C(130399)

General description

Activin receptor type-1 (ACVR1C) is a dimeric growth and differentiation factor belonging to the TKL Ser/Thr protein kinase family. Activins signal through a heteromeric complex of receptor serine kinases which include at least two, type I and two type II receptors. ACVR1C plays an important role in cell differentiation, growth arrest, and apoptosis. It is found in pancreas, heart, colon, small intestine, ovary, hippocampus, medulla oblongata, and putamen of the brain.

Immunogen

KLH-conjugated linear peptide corresponding to human ACVR1C.

Application

Detect Activin receptor type-1 using this rabbit polyclonal antibody, Anti-ACVR1C Antibody validated for use in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in human placenta tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected ACVR1C in 15 µg of human placenta tissue lysate.

Target description

~62 kDa observed. Uniprot describes 4 isoforms at ~37 kDa, ~46 kDa, ~50 kDa, and ~55 kDa Uncharacterized bands may appear in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human placenta tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service