Skip to Content
MilliporeSigma
All Photos(3)

Documents

ABN466

Sigma-Aldrich

Anti-NANOS2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nanos homolog 2, NOS-2, NANOS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NANOS2(339345)

General description

Nanos homolog 2 (NANOS2) is a zinc-finger containing, RNA-binding protein belonging to the nanos family. NANOS2 plays an important role in the sexual differentiation of germ cells by promoting the male fate but suppressing the female fate. It has been implicated in germ cell development in both invertebrates and vertebrates. Unlike NANOS1, whose expression in mice is dispensable, the NANOS2 and NANOS3 proteins are required for germ cell development. NANOS2-null primordial germ cells (PGCs) die only in the male gonads and show no defects in females, while NANOS3-null PGCs are lost during the migration stage regardless of sex.

Specificity

This antibody recognizes the N-terminus of NANOS2.

Immunogen

KLH-conjugated linear peptide corresponding to the N-terminus of human NANOS2.

Application

Detect Nanos homolog 2 using this rabbit polyclonal antibody, Anti-NANOS2 Antibody validated for use in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: 2.5 µg/mL from a representative lot detected NANOS2 in human brain tissue.

Immunofluorescence Analysis: 20 µg/mL from a representative lot detected NANOS2 in human testis cells.

Quality

Evaluated by Western Blotting in rat brain tissue.

Western Blotting Analysis: 2 µg/mL of this antibody detected NANOS2 in 15 µg of rat brain tissue.

Target description

~15 kDa observed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service