Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN404

Sigma-Aldrich

Anti-MCPH1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Microcephalin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$454.00

$454.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$454.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$454.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MCPH1(79648)

General description

The protein named Microcephalin and encoded by the human gene named MCPH1 is a DNA/chromatin binding protein implicated in chromosome condensation and DNA damage induced cellular responses. MCPH1 plays a role in neurogenesis and is involved in the regulation of the size of the cerebral cortex. MCPH1 interacts with several proteins including H2AFX under DNA damage conditions and CDC27 during normal cell growth. MCPH1 is expressed cytoplasmically in a variety of cell types and tissues and is particularly expressed in the fetal brain, liver and kidney. Defects in MCPH1 can give rise to microcephaly a disease where the head circumference is markedly reduced and effected patients are mentally retarded, showing a high number of prophase-like cells in cytological examinations. In addition, cell divisions are irregular and demonstrate a delay in post-mitotic chromosome decondensation. Interestingly, recent research demonstrates that MCPH1 also functions as a tumor suppressor that is regulated by the microRNA miR-27a.

Immunogen

KLH-conjugated linear peptide corresponding to human MCPH1.

Application

Anti-MCPH1 Antibody is a highly specific rabbit polyclonal antibody, that targets Microcephalin & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: A 1:500-2,000 dilution from a representative lot detected MCPH1 in human cerebellum, human testis, and human cerebral cortex tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in human testis tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MCPH1 in 10 µg of human testis tissue lysate.

Target description

~111 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service