Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN2181

Sigma-Aldrich

Anti-NCAM-L1

from rabbit

Synonym(s):

Neural cell adhesion molecule L1-like protein, L1 adhesion molecule, Cell adhesion molecule with homology to L1CAM, Chl1-like protein, Close homolog of L1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$607.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$607.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Chl1(12661)

General description

Neural cell adhesion molecule L1-like protein (UniProt: P70232; also known as Cell adhesion molecule with homology to L1CAM, NCAM-L1, Chl1-like protein, Close homolog of L1) is encoded by the Chl1 (also known as Call) gene (Gene ID: in murine species. NCAM-L1 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off in the mature form. It is expressed in neurons and glial cells in the brain and the spinal cord and has also been detected in the retina and the optic nerve. It is not detectable in mouse forebrain at E11 and becomes weakly detectable at E13 with highest detection at E18 to postnatal day 7. It is down-regulated at postnatal day 15 and is further reduced in four-week-old animals. NCAM-L1 is heavily N-glycosylated and contains N-linked oligosaccharides with a sulfated carbohydrate structure type HNK-1. NCAM-L1 contains an extracellular domain (aa 26-1083), a transmembrane domain (aa 1084-1104), and a cytoplasmic domain (aa 1105-1209). It also has six Ig-like C2 type domains and four fibronectin type -III domains. NCAM-L1 is an extracellular matrix and cell adhesion protein that plays a role in nervous system development and in synaptic plasticity. It plays a role in neuronal positioning of pyramidal neurons as well as in regulation of both the number of interneurons and the efficacy of GABAergic synapses.

Specificity

This rabbit polyclonal antibody detects NCAM-L1 protein in mouse and rat.

Immunogen

Epitope: unknown
Purified full-length murine Neural cell adhesion molecule L1-like protein (NCAM-L1).

Application

Anti-NCAM-L1, Cat. No. ABN2181, is a highly specific rabbit polyclonal antibody that targets Neural cell adhesion molecule L1 and has been tested in Immunofluorescence, Immunoprecipitation, and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected NCAM-L1 in Western Blotting applications (Schmidt, H., et. al. (2002). J Cell Biol. 159(3):489-98; Alberts, P., et. al. (2003). Mol Biol Cell. 14(10):4207-20).

Immunoprecipitation Analysis: A representative lot immunoprecipitated NCAM-L1 in Immunoprecipitation applications (Patel, K., et. al. (1991). Hybridoma. 10(4):481-91).

Western Blotting Analysis: 0.5 µg/mL from a representative lot detected NCAM-L1 in crude membrane fractions of postnatal day 10 brain extracts (Courtesy of Fritz G. Rathjen from Max-Delbrück-Center, Berlin Germany).

Immunofluorescence Analysis: A representative lot detected NCAM-L1 in Immunofluorescence applications (Ter-Avetisyan, G., et. al. (2014). J Neurosci. 34(3):737-47; Abe, P., et. al. (2015). J Neurosci. 35(38):13053-63).

Quality

Evaluated by Western Blotting in Wild type & knock out P10 mouse brain extract crude membrane.

Western Blotting Analysis: 2 µg/mL of this antibody detected NCAM-L1 in crude membrane extact from brain of Wild type, but not in NCAM-L1 knock out P10 mouse.

Target description

~220 kDa observed; 135.07 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service