Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABN1734

Sigma-Aldrich

Anti-SNAT2

from rabbit

Synonym(s):

Sodium-coupled neutral amino acid transporter 2, Amino acid transporter A2, Solute carrier family 38 member 2, System A amino acid transporter 2, System A transporter 1, System N amino acid transporter 2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$231.00
100 μL
$517.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$231.00
100 μL
$517.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Sodium-coupled neutral amino acid transporter 2 (UniProt: Q9JHE5; also known as Amino acid transporter A2, Solute carrier family 38 member 2, System A amino acid transporter 2, System A transporter 1, System N amino acid transporter 2) is encoded by the Scl38a2 (also known as Ata2, Sa1, Sat2, Snat2) gene (Gene ID: 29642) in rat. Sodium-coupled neutral amino acid transporter 2 is a multi-pass membrane protein that functions as a sodium-dependent amino acid transporter and mediates the saturable, pH-sensitive and electrogenic cotransport of neutral amino acids and sodium ions with a stoichiometry of 1:1. It may also function in the transport of amino acids at the blood-brain barrier and in the supply of maternal nutrients to the fetus through the placenta. Its transport action is reported to be inhibited by N-methyl-D-glucamine and choline. It is enriched in the somatodendritic compartment of neurons and is also detected at the axonal shaft, but is excluded from the nerve terminal. It is also expressed in skeletal muscle and adipose tissue. Its levels are up-regulated upon amino acid deprivation that results from increased transcription and protein stabilization.

Specificity

This polyclonal antibody detects sodium-coupled neutral amino acid transporter 2 (SNAT2) in rat brain. It targets an epitope within the first 65 amino acids.

Immunogen

Epitope: N-terminus
GST-tagged recombinant fragment corresponding to the first 65 amino acids from te N-terminal region of rat Sodium-coupled neutral amino acid transporter 2 (SNAT2).

Application

Anti-SNAT2, Cat. No. ABN1734, is a highly specific rabbit polyclonal antibody that targets Sodium-coupled neutral amino acid transporter 2 and has been tested in Immunocytochemistry, Immunofluorescence, Immunohistochemistry, and Western Blotting.
Immunofluorescence Analysis: A 1:150 dilution from a representative lot detected SNAT2 in rat cerebral cortex and human cerebellum tissue sections.

Immunocytochemistry Analysis: A representative lot detected SNAT2 in Immunocytochemistry applications (Grewal, S., et. al. (2009). J Biol Chem. 284(17):11224-36).

Western Blotting Analysis: A representative lot detected SNAT2 in Western Blotting applications (Yao, D., et. al. (2000). J Biol Chem. 275(30):22790-7; Melone, M., et. al. (2006). Neuroscience. 140(1):281-92; Grewal, S., et. al. (2009). J Biol Chem. 284(17):11224-36).

Immunohistochemistry Analysis: A representative lot detected SNAT2 in Immunohistochemistry applications (Melone, M., et. al. (2006). Neuroscience. 140(1):281-92).
Research Category
Neuroscience

Quality

Evaluated by Immunofluorescence in rat brain sections.

Immunofluorescence Analysis: A 1:150 dilution of this antibody detected SNAT2 in rat brain tissue sections.

Target description

55.55 kDa calculated.

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.05% sodium azide.
Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Stable for 1 year at -20°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service