Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABF215

Sigma-Aldrich

Anti-UNC93B1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

UNC93B1 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$446.00

$446.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$446.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UNC93B1(81622)

General description

UNC93B1 protein (UNC93B1) is a human homolog of the C. elegans Unc93 gene. UNC93B1 specifically interacts with TLR3, 7 and 9; the histidine-to-arginine point mutation used to identify Unc93b abolishes this interaction. UNC93B1 may play an important role in innate immunity.

Immunogen

KLH-conjugated linear peptide corresponding to human UNC93B1.

Application

Detect UNC93B1 protein using this rabbit polyclonal antibody, Anti-UNC93B1 Antibody validated for use in western blotting, IHC (Paraffin) & Immunofluorescence.
Immunohistochemistry Analysis: 2.5 µg/mL from a representative lot detected UNC93B1 in human heart tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected UNC93B1 in human heart tissue.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blotting in human heart tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected UNC93B1 in 15 µg of human heart tissue lysate.

Target description

~65 kDa observed. Uncharacterized bands may appear in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human heart tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service