Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABF1067M

Sigma-Aldrich

Anti-Tapasin

from rabbit

Synonym(s):

Tapasin, TPN, TPSN, TAP-associated protein, TAP-binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$231.00
100 μL
$514.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$231.00
100 μL
$514.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

packaging

antibody small pack of 25 μL

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Tapbp(21356)

Related Categories

General description

Tapasin (UniProt: Q9R233; also known as TPN, TPSN, TAP-associated protein, TAP-binding protein) is encoded by the Tapbp (also known as Tapa) gene (Gene ID: 21356) in the murine species. Tapasin is a single-pass type I membrane glycoprotein protein that is involved in the association of MHC class I with transporter associated with antigen processing (TAP) and in the assembly of MHC class I with peptide. It is synthesized with a signal peptide of 23 amino acids, which is subsequently cleaved to generate mature Tapasin. It assists in loading of antigenic peptides on to class I molecules in the endoplasmic reticulum (ER). It is also reported to support the binding of high-affinity peptides to MHC class I in conjunction with ERp57. Defects in Tapasin expression are linked to destabilization of the MHC class I loading complex and diminution in the expression of MHC molecules at the cell surface. Tapasin is reportedly down-regulated in several human carcinomas and its re-expression in Tapasin-deficient cancer cells restores the expression of MHC class I antigen complexes and augment cancer cell immunogenicity, which helps in longer survival in animals bearing metastatic tumors. (Ref.: Lou, Y et al. (2008). Clin Cancer Res. 14(5); 1494-1501).

Specificity

This rabbit polyclonal antibody detects Tapasin in murine cells. It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of murine Tapasin.

Application

Anti-Tapasin Antibody, Cat. No. ABF1067, is a rabbit polyclonal antibody that detects Tapasin in murine cells and has been tested for use in Immunoprecipitation and Western Blotting.
Immunoprecipitation Analysis: A representative lot detected Tapasin in K42 cells (Lum, R., et. al. (2016). J Biol Chem. 291(37):19631-41) and L-cells and BW5147 thymoma cells (Suh, W.K., et. al. (1999). J Immunol. 162(3):1530-40).

Western Blotting Analysis: A representative lot detected Tapasin in CMT.64 cells treated with IFN- (Lou, Y., et. al. (2008). Clin Cancer Res. 14(5):1494-501).
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Tapasin in 10 µg of NIH/3T3 cell lysate.

Target description

~51 kDa observed; 49.74 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service