Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE81

Sigma-Aldrich

Anti-NXF1 Antibody

serum, from rabbit

Synonym(s):

Nuclear RNA export factor 1, Tip-associated protein, Tip-associating protein, mRNA export factor TAP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$376.00

$376.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$376.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$376.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NXF1(10482)

General description

Nuclear RNA export factor 1 (also known as Tap protein) is encoded by the NXF1 gene. The protein expressed by this gene is one member of a family of nuclear RNA export factor gene. The Nuclear RNA export factor 1 protein is involved in export of mRNA species bearing retroviral constitutive transport elements (CTE) from the nucleus as well as in the export of mRNA from the nucleus to the cytoplasm. This protein is localized predominantly in the nucleoplasm at both the nucleoplasmic and cytoplasmic faces of the nuclear pore complex. However, this protein shuttles between the nucleus and the cytoplasm and binds in vivo to poly(A) RNA. This protein is present in the nuclear Y14 complex and has been shown to interact with Aly/REF.

Immunogen

Recombinant protein corresponding to human NXF1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-NXF1 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect NXF1 also known as Nuclear RNA export factor 1, Tip-associated protein.

Quality

Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected NXF1 on 10 µg of A431 cell lysate.

Target description

~72 kDa observed. Two isoforms at 70 and 40 kDa have been described. Uncharacterized bands at ~48 and 100 kDa have been observed in some lysates.

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service