Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE606

Sigma-Aldrich

Anti-PERP Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

p53 apoptosis effector related to PMP-22, PERP, Keratinocyte-associated protein 1, KCP-1, Krtcap1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PERP(64065)

General description

P53 apoptosis effector related to PMP-22 (PERP), also known as Keratinocyte-associated protein 1 (KCP-1, Krtcap1), is a member of the TMEM47 family and a component of the intercellular desmosome junctions. PERP has been shown to play an important role in the stratified epithelial integrity and cell-cell adhesion by promoting desmosome assembly. Recent evidence implies that PERP is a mediator of p53 induced apoptosis. P53 binds to the promoter of PERP and activates the PERP gene and consequently the translated PERP protein mediates the p53 induced apoptosis. PERP is expressed in apoptotic cells and in the stratified squamous skin epithelium of the skin and the tongue, but not in simple epithelia.

Immunogen

KLH-conjugated linear peptide corresponding to human PERP.

Application

Detect P53 apoptosis effector related to PMP-22 using this rabbit polyclonal antibody, Anti-PERP Antibody validated for use in western blotting & ICC.
Immunocytochemistry Analysis: 10 µg/mL from a representative lot detected PERP in A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blotting/Peptide Inhibition Assay in A431 cell lysate.

Western Blotting/Peptide Inhibition Assay Analysis: 1 µg/mL of this antibody detected PERP in 15 µg of A431 cell lysate.

Target description

~21 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service