Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE598

Sigma-Aldrich

Anti-TCF12 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

HTF4, HEB, BHLHB20, Transcription factor 12, TCF-12, Class B basic helix-loop-helix protein 20, bHLHb20, DNA-binding protein HTF4, E-box-binding protein, Transcription factor HTF-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TCF12(6938)

General description

TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family. TCF12 is a transcriptional regulator involved in the initiation of neuronal differentiation, and it has been shown to activate transcription by binding to the E box (5′-CANNTG-3′). TCF12 has been shown to interact with PTF1A, NEUROD2 and RUNX1T1. TCF12 is suggested to play a critical role in proper T cell differentiation by blocking the thymocyte proliferation in combination with E2A. Recent studies have shown that TCF12 is also an important factor required for the development of invariant natural killer T cells. TCF12 is expressed in many tissues, and more specifically in skeletal muscle, thymus, B- and T-cells.

Immunogen

KLH-conjugated linear peptide corresponding to human TCF12.

Application

Detect TCF12 using this rabbit polyclonal antibody, Anti-TCF12 Antibody validated for use in western blotting, ICC & IHC.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected TCF12 in human tonsil tissue.

Immunocytochemistry Analysis: 10 µg/mL from a representative lot detected TCF12 in HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected TCF12 in 15 µg of HeLa cell lysate.

Target description

~83 kDa observed. Uncharacterized bands may appear in some lysates. Uniprot describes 3 isoforms between ~55 kDa and ~76 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service