Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE499

Sigma-Aldrich

Anti-JMJD2D Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Lysine-specific demethylase 4D, JmjC domain-containing histone demethylation protein 3D, Jumonji domain-containing protein 2D

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$485.00

$485.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$485.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$485.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... KDM4D(55693)

General description

Jumonji domain-containing protein 2D (JMJD2D) is also called Lysine-specific demethylase 4D and JmjC domain-containing histone demethylation protein 3D. JMJD2D is a histone demethylase that specifically demethylates both di- and trimethylated H3 ′Lys-9′, but not monomethylated histone H3 ′Lys-9′ residues. JMJD2D also does not demethylate histone H3 ′Lys-4′, H3 ′Lys-27′, H3 ′Lys-36′ or H4 ′Lys-20′.

Specificity

Other homologies: Bovine, Porcine, Canine, and Rhesus Macaque (93% sequence homology), Mouse (87% sequence homology), Rat (80% sequence homology).

Immunogen

KLH-conjugated linear peptide corresponding to human JMJD2D.

Application

Detect Jumonji domain-containing protein (JMJD) using this rabbit polyclonal antibody, Anti-JMJD2D Antibody validated for use in western blotting, IHC & IP.
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected JMJD2D in human testis and liver tissues.

Immunoprecipitation Analyis: 20 µg from a representative lot detected JMJD2D in 500 µg of HeLa cell lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected JMJD2D in 10 µg of HeLa cell lysate

Target description

~58 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service