Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE421

Sigma-Aldrich

Anti-GATA-like protein 1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

GATA-type zinc finger protein 1, GATA-like protein 1, GLP-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$564.00

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$564.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

GATA-like protein 1 (GLP-1) is also known as GATA-type zinc finger protein 1. GATA-like protein 1 is a zinc finger protein that functions as a transcriptional repressor. GATA-like protein 1 represses GATA transcription factor function, is required for germ cell development and is crucial for somatic cells of the gonad.

Specificity

This antibody recognizes GATA -like protein 1 near the N-terminus.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human GATA-like protein 1 near the N-terminus.

Application

Anti-GATA-like protein 1 Antibody is a highly specific rabbit polyclonal antibody, that targets GATA-like protein 1 & has been tested in western blotting & IHC (Paraffin).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected GATA-like protein 1 in 10 µg of human prostate tissue lysate.

Quality

Evaluated by Immunohistochemistry in human colon tissue.

Imunnohistochemistry Analysis: A 1:1,000 dilution of this antibody detected GATA-like protein 1 in human colon tissue.

Target description

~32 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human colon tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Kruse et al.
Arteriosclerosis, thrombosis, and vascular biology, 35(9), 1954-1962 (2015-07-25)
The polarity protein Scrib is highly expressed in endothelial cells and is required for planar cell polarity. Scrib also facilitates recycling of integrin α5 to the plasma membrane. Because integrin α5 signals the presence of the inflammatory matrix protein fibronectin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service