Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE409

Sigma-Aldrich

Anti-UTX Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Lysine-specific demethylase 6A, Histone demethylase UTX, Ubiquitously-transcribed TPR protein on the X chromosome, Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$564.00

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$564.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM6A(7403)

General description

Histone demethylase UTX is also known as Lysine-specific demethylase 6A, Ubiquitously-transcribed TPR protein on the X chromosome, and Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein. UTX is a histone demethylase that specifically demethylates trimethylated and dimethylated but not monomethylated histone H3 ′Lys-27′. UTX is involved in regulation of posterior development, and defects can cause Kabuki syndrome type 2 (KABUK2)

Specificity

Other homologies: Canine (99% sequence homology) and Bovine (96% sequence homology).

Immunogen

GST-tagged recombinant protein corresponding to human UTX.

Application

Detect Histone demethylase UTX using this rabbit polyclonal antibody, Anti-UTX Antibody validated for use in western blotting & ICC.
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected UTX in HeLa, A431, and 3T3 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected UTX in 10 µg of HeLa nuclear extract.

Target description

~154 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenyu Wang et al.
Molecular cancer therapeutics, 17(9), 1973-1983 (2018-06-22)
Despite showing promise against PIK3CA-mutant breast cancers in preclinical studies, PI3K/AKT pathway inhibitors demonstrate limited clinical efficacy as monotherapy. Here, we found that histone H3K27me3 demethylase KDM6B-targeted IGFBP5 expression provides a protective mechanism for PI3K/AKT inhibitor-induced apoptosis in breast cancer
Xuepei Lei et al.
Stem cell reports, 10(4), 1193-1207 (2018-03-20)
Neural stem cell (NSC) proliferation and differentiation in the developing brain is a complex process precisely regulated by intrinsic and extrinsic signals. Although epigenetic modification has been reportedly involved in the regulation of the cerebral cortex, whether UTX, an H3K27me3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service