Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE386

Sigma-Aldrich

Anti-Flightless-1 homolog Antibody

serum, from rabbit

Synonym(s):

Protein flightless-1 homolog, Fli1, Anti-Flightless-1 homolog Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$359.00

$359.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$359.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$359.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FLII(2314)

General description

Mammalian homolog of drosophila flightless-1 is encoded by the FLII gene in human (also known as FLI, Fli1, or FLIL). The human flightless-1 homolog (Flii) contains sixteen 23-amino acid leucine-rich repeats (LRR) in its N-terminal half and a gelsolin-like domain (GLD) in its C-terminal half, composed of six segmental repeats typical of many gelsolin family members. Flii is reported to interact with actin and "FLI LRR associated protein" (FLAP) via its GLD and LRR domain, respectively (PMID 9525888). Flii is also shown to interact with the glucose-responsive transcription factor carbohydrate responsive element binding protein (ChREBP) via both the LRR and GLD domains and functions as a negative regulator of ChREBP in cancer cells (PMID 24055811). Increased Flii level is observed in the blistered skin of patients with epidermolysis bullosa acquisita (EBA), and Flii overexpression in transgenic mice results in delayed epidermal barrier formation in embryos and decreased expression of tight junction (TJ) proteins, as well as impaired barrier function among adult transgenic mice (PMID 24375017).

Immunogen

KLH-conjugated linear peptide corresponding to human Flightless-1 homolog.

Application

Detect Fli1 using this rabbit polyclonal antibody, Anti-Flightless-1 homolog Antibody validated for use in western blotting & IP.
Immunoprecipitation Analysis: A representative lot of this antibody detected Flightless-1 homolog in IP (Prof. D. Pallas, Emory University).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Flightless-1 homolog in 10 µg of A431 cell lysate.

Target description

~144 kDa observed

Linkage

Replaces: MABF195; MABE530

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service