Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

ABE2922

Sigma-Aldrich

Anti-Histone H1E frameshift mutant

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$239.00
100 μG
$603.00

$239.00


Please contact Customer Service for Availability


Select a Size

Change View
25 μG
$239.00
100 μG
$603.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$239.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 21.86 kDa
observed mol wt ~33 kDa

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

affinity binding assay: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

mutation (Frameshift aa145-150)

Gene Information

human ... Unknown(3008)

General description

Histone 1, H1e (UniProt: A3R0T8; also known as Histone H1.4, HIST1H1E) is encoded by the HIST1H1E (also known as hCG_16457421) gene (Gene ID: 3008) in human. Histones have been classified under five major classes. The core histones H2A, H2B, H3, and H4 constitute the protein components of nucleosome core particles and histone H1 is a linker histone that plays a role in chromatin folding. Depletion of histone H1 can cause dramatic changes in chromatin structure, global nucleosome spacing, and reduced chromatin compaction. Pathogenic mutations in HIST1H1E gene result in the formation of histone H1 that is less effective in neutralizing negatively charged linker DNA and display reduced DNA binding and protein-protein interactions due to truncation in key residues. These mutations are reported to result in a shift in reading frame in the C-terminal region and produce truncated proteins. Frameshift mutations in HIST1H1E have been linked to Rahman syndrome that is manifested in overgrowth with intellectual disabilities (OGID) in early infancy. Subjects with Rahman syndrome display a global tendency in reduction of methylation state. (Ref.: Andrea Ciolfi, et al. (2020). Clinical Epigenetics 12; Article 7; Tatton-Brown , K., et al. (2017). Am. J. Hum. Genet. 100(5); 725-736).

Specificity

This rabbit polyclonal antibody specifically detects Histone H1 with H1E frameshift mutation.

Immunogen

KLH-conjugated Linear Peptide corresponding to the Unknown of Human Histone H1.

Application

Quality Control Testing

Evaluated by Western Blotting in human oligodendroglioma cell line SCC163 stably expressing transgenic H1Efs-FLAG-HA.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Histone H1E frameshift mutant in human oligodendroglioma cell line SCC163 stably expressing transgenic H1Efs-FLAG-HA.

Tested Applications

Immunofluorescence Analysis: A representative lot detected Histone H1E frameshift mutant in mESc and H1Eframeshift Knock-in cells (Courtesy of Prof. C. David Allis Lab at Rockefeller University).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Histone H1E frameshift mutant in Human WBC and mESC′s & various other cell lines (Courtesy of Prof. C. David Allis Lab at Rockefeller University).

Affinity Binding Assay: A representative lot of this antibody bound Histone H1E peptide with a KD of 1.0 x 10-12

in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Histone H1E frameshift mutant, Cat. No. ABE2922, is a rabbit polyclonal antibody that detects Histone H1E frameshift mutant and is tested for use in Affinity Binding Assay, Immunofluorescence, and Western Blotting.

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service