Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE2873

Sigma-Aldrich

Anti-NUDT21

from rabbit

Synonym(s):

Cleavage and polyadenylation specificity factor subunit 5, Cleavage and polyadenylation specificity factor 25 kDa subunit, CPSF 25 kDa subunit, Nucleoside diphosphate-linked moiety X motif 21, Nudix motif 21, Pre-mRNA cleavage factor Im 25 kDa subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... NUDT21(11051)

General description

Cleavage and polyadenylation specificity factor subunit 5 (UniProt: O43809; also known as Cleavage and polyadenylation specificity factor 25 kDa subunit, CFIm25, CPSF 25 kDa subunit, Nucleoside diphosphate-linked moiety X motif 21, Nudix motif 21, Pre-mRNA cleavage factor Im 25 kDa subunit) is encoded by the NUDT21 (also known as CFIM25, CPSF25, CPSF5) gene (Gene ID: 11051) in human. CFlm25 is a homodimeric protein that is a component of the cleavage factor Im (CFIm) complex, which plays a key role in pre-mRNA 3′-processing. It is involved in association with CPSF6 or CPSF7 in pre-MRNA 3′-end poly(A) site cleavage and poly(A) addition. The interaction of CFlm25 with the RNA is one of the earliest steps in the assembly of the 3′ end processing complex and facilitates the recruitment of other processing factors. It contains one Nudix hydrolase domain (aa 76-201). The region containing aa 2-147 is considered to be essential for RNA-binding. It can be acetylated by p300/CBP and acetylation results in a reduction in its interaction with Poly(A) polymerase alpha (PAPAO).

Specificity

This rabbit polyclonal antibody detects Cleavage and polyadenylation specificity factor subunit 5 (CFIm25) in human cells.

Immunogen

Epitope: unknown
His-tagged full length recombinant human Cleavage and polyadenylation specificity factor subunit 5 (CFIm25).

Application

Anti-NUDT21, Cat. No. ABE2873, is a highly specific rabbit polyclonal antibody that targets Cleavage and polyadenylation specificity factor subunit 5 and has been tested in Immunoprecipitation and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected NUDT21 in RNase A-treated HeLa whole-cell extract after IP (Katahira, J., et. al. (2013). Nucleic Acid Res. .41(14):7060-72).

Immunoprecipitation Analysis: A representative lot detected NUDT21 in Co-immunoprecipitation with FLAG-tagged CF I cell lysates (Shimazu, T., et. al. (2007). J Biol Chem. 282(7):4470-8).

Quality

Evaluated by Western Blotting in Caco-2 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected NUDT21 in 10 µg of Caco-2 cell lysate.

Target description

~25 kDa observed; 26.23 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service