Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE2619

Sigma-Aldrich

Anti-Polo-like kinase-1

from rabbit

Synonym(s):

Serine/threonine-protein kinase PLK1, EC: 2.7.11.21, PLK-1, Serine/threonine-protein kinase 13, STPK13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PLK1(5347)

General description

Serine/threonine-protein kinase PLK1 (UniProt: P53350; also known as EC: 2.7.11.21, Polo-like kinase 1, PLK-1, Serine/threonine-protein kinase 13, STPK13) is encoded by the PLK1 (also known as PLK) gene (Gene ID: 5347) in human. PLK-1 is a serine/threonine-protein kinase that performs several important functions throughout M phase of the cell cycle, including the regulation of centrosome maturation and spindle assembly, the removal of cohesins from chromosome arms, the inactivation of anaphase-promoting complex/cyclosome (APC/C) inhibitors, and the regulation of mitotic exit and cytokinesis. It is shown to express in placenta and colon and its activity is enhanced by growth promoting agents. PLK-1 is activated by phosphorylation at Thr210 by aurora kinase A. It accumulates to a maximum during the G2 and M phases and declines to a nearly undetectable levels following mitosis and throughout G1 phase. However, it begins to accumulate again during the S phase of cell cycle. The kinase domain of PLK-1 is localized to amino acids 53-305 with activation loop in the region of amino acids 194-221. PLK-1 contains 2 POLO box domains (aa 417-480 and 515-584), which act as phosphopeptide-binding module that recognize and bind serine-phosphothreonine/ phosphoserine]-(proline/X) motifs. PLK-1 recognizes and binds docking proteins that are already phosphorylated on these motifs, and then phosphorylates them. Defects in PLK1 gene are associated with some cancers, such as gastric, thyroid or B-cell lymphomas. Expression is shown to be higher in tumor tissues with a poor prognosis, suggesting its role in malignant transformations and carcinogenesis.

Specificity

This rabbit polyclonal antibody detects human Polo-like kinase 1. It targets an epitope within 303 amino acids from the C-terminal half.

Immunogen

Epitope: C-terminus
His-tagged recombinant fragment corresponding to 303 amino acids from the C-terminal half of human Polo-like kinase-1.

Application

Anti-Polo-like kinase-1, Cat. No. ABE2619, is a rabbit polyclonal antibody that detects Polo-Like Kinse 1 and has been tested for use Immunocytochemistry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected Polo-like kinase-1 in U2OS cells (Macurek, L., et. al. (2008). Nature. 455(7209):119-23).

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Polo-like kinase-1 in U2OS and C2C12 cells.

Western Blotting Analysis: A representative lot detected Polo-like kinase-1 in U2OS-TR transfected cells (Macurek, L., et. al. (2008). Nature. 455(7209):119-23).

Immunoprecipitation Analysis: A representative lot detected Polo-like kinase-1 in U2OS-TR transfected cells (Macurek, L., et. al. (2008). Nature. 455(7209):119-23).
Research Category
Signaling

Quality

Evaluated by Western Blotting in U2OS transfected with PLK1 siRNA and U2OS transfected with control oligo (LUC o Luciferase).

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Polo-like kinase-1 in U2OS transfected with PLK1 siRNA (PLK1) & U2OS transfected with control oligo (LUC o Luciferase).

Target description

~68 kDa observed; 68.26 kD calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS containing 80 mM Glycine with 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service