Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE1958

Sigma-Aldrich

Anti-Zic1/2

from rabbit

Synonym(s):

Zinc finger protein of the cerebellum 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$516.00

$516.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$516.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$516.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Zic1(22771)

General description

Zinc finger protein ZIC 1 (UniProt: P46684; also known as Zinc finger protein of the cerebellum 1) is encoded by the Zic 1 (also known as Zic) gene (Gene ID: 22771) in murine species. Zic 1 is a member of the GLI C2H2-type zinc-finger protein family. Zic 1 is expressed in the central nervous system and in osteoblasts. A high level expression is seen in the cerebellum, while a low level expression is seen in the olfactory bulb, diencephalon, and brainstem. It acts as a transcriptional activator that is involved in neurogenesis and plays important roles in the early stage of organogenesis of the CNS, as well as during dorsal spinal cord development and maturation of the cerebellum. The C2H2-type 3, 4 and 5 zinc finger domains of Zic 1 are considered to be essential for transcription activation and for Zic 1 interaction with MyoD family inhibitor domain-containing (MDFIC) protein. Zic 1 is also shown to be involved in the spatial distribution of mossy fiber (MF) neurons within the pontine gray nucleus (PGN). It plays a role in the regulation of MF axon pathway choice and promotes MF migration towards ipsilaterally-located cerebellar territories.

Specificity

This polyclonal antibody detects Zic1 protein in mouse and human.

Immunogen

A linear peptide corresponding to 20 amino acids from the C-terminal end of mouse Zic 1 protein.
Epitope: C-terminus

Application

Anti-Zic1/2, Cat. No. ABE1958, is a highly specific rabbit polyclonal antibody that targets Zinc finger protein ZIC 1 and has been tested in Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Immunofluorescence, and Western Blotting.
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Zic1/2 in mouse cerebellum (Frank, C.L., et. al. (2015). Nat Neurosci. 18(5):647-56).

Immunofluorescence Analysis: A representative lot detected Zic1/2 in postmitotic granule cells (Kim, J.Y., et. al. (2003). Dev Biol. 263(1):50-66) and adult mouse cerebellum (Koirala, S., et. al. (2009). J Neurosci. 29(23):7439-49).

Immunocytochemistry Analysis: A representative lot detected Zic1/2 in Immunocytochemistry applications (Kenney, A.M., et. al. (2000). Mol Cell Biol. 20(23):9055-67).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in human cerebellum tissue lysate.

Western Blotting Analysis: 1:500 dilution of this antibody detected Zic1/2 in 10 µg of human cerebellum tissue lysate.

Target description

~53 kDa observed; 48.33 kDa calculated for Zic1; 54.95 kDa calculated for Zic2. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service