Skip to Content
MilliporeSigma
All Photos(2)

Documents

ABE1797

Sigma-Aldrich

Anti-Transcription Initiation Factor IIA subunit 1

from rabbit, purified by affinity chromatography

Synonym(s):

General transcription factor IIA subunit 1, TFIIAL, Transcription initiation factor TFIIA 42 kDa subunit, TFIIA-42

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GTF2A1(2957)

General description

Transcription initiation factor IIA subunit 1 (UniProt: P52655; also known as General transcription factor IIA subunit 1, TFIIAL, Transcription initiation factor TFIIA 42 kDa subunit, TFIIA-42) is encoded by the GTF2A1 (also known as TF2A1) gene (Gene ID: 2957) in human. TFIIA is one of the several components of the transcription machinery of RNA polymerase II that plays an important role in transcriptional activation. TFIIA in a complex with TATA-box-binding protein (TBP) mediates transcriptional activity. After initiation, some components of the pre-initiation complex, including TFIIA, remain attached and re-initiate a subsequent round of transcription. Binding of TFIIA to TBP helps to stabilize TBP binding to the TATA element. TFIIA is a heterodimer of the large unprocessed subunit 1 and a small subunit gamma. It was originally believed to be a heterotrimer of an alpha (p35), a beta (p19) and a gamma subunit (p12).

Specificity

This rabbit polyclonal antibody detects Transcription initiation factor IIA subunit 1 in human cells. It targets an epitope within 15 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Transcription initiation factor IIA subunit 1.

Application

Anti-Transcription Initiation Factor IIA subunit 1, Cat. No. ABE1797, is a highly specific rabbit polyclonal antibody that targets Transcription initiation factor IIA subunit 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 0.2 µg/mL from a representative lot detected Transcription initiation factor IIA subunit 1 in 10 µg of RAW264.7 cell lysate.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Transcription initiation factor IIA subunit 1 in human brain and human testis tissue.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Transcription initiation factor IIA subunit 1 in 10 µg of HeLa cell lysate.

Target description

~55 kDa observed; 41.51 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service