Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE1420

Sigma-Aldrich

Anti-Mediator complex subunit 25

from rabbit, purified by affinity chromatography

Synonym(s):

Mediator of RNA polymerase II transcription subunit 25, Activator interaction domain-containing protein 1, Activator-recruited cofactor 92 kDa component, ARC92

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$475.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$475.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), yeast (based on 100% sequence homology), bovine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MED25(81857)

General description

Mediator of RNA polymerase II transcription subunit 25 (UniProt: Q71SY5; also known as Activator interaction domain-containing protein 1, Activator-recruited cofactor 92 kDa component, ARC92, Mediator complex subunit 25, p78) is encoded by the MED25 (also known as ACID1, ARC92, PTOV2, TCBAP0758) gene (Gene ID: 81857) in human. It is a member of the Mediator complex subunit 25 family that is ubiquitously expressed. Higher expression levels are observed in brain, heart, kidney, peripheral leukocytes, placenta, skeletal muscle and spleen. It acts as a coactivator that is involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator complex subunit 25 functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. It is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors. Six isoforms of Mediator complex subunit 25 have been described that are generated by alternative splicing. Defects in MED25 gene are known to cause Charcot-Marie-Tooth disease 2B2, a disorder of the peripheral nervous system characterized by progressive weakness and atrophy.

Specificity

This rabbit polyclonal antibody detects Mediator of RNA polymerase II transcription subunit 25 (MED25) in human and murine species. it targets an epitope within 16 amino acids from the internal region. Detects all isoforms of Mediator complex subunit 25.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the internal region of human Mediator of RNA polymerase II transcription subunit 25 (MED25).

Application

Anti-Mediator complex subunit 25, Cat. No. ABE1420, is a highly specific rabbit polyclonal antibody that targets MED25 and has been tested in Immunohistochemistry (Paraffin), Peptide Inhibition Assay, and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Mediator complex subunit 25 in human cerebellum and mouse cerebral cortex tissues.

Peptide Inhibition Analysis: 2 µg/mL from a representative lot blocked Mediator complex subunit 25 in mouse brain tissue lysate.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Mediator complex subunit 25 in 10 µg of HeLa cell lysate.

Target description

~70 kda observed; 78.17 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine, 0.15 M NaCl, pH 7.4 with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service